Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596395_m1
          See other PPP1R17 GE Assays ›
          Gene Symbol:
          Ppp1r17
          Gene Name:
          protein phosphatase 1, regulatory subunit 17
          Gene Aliases:
          Gsbs
          Chromosome Location:
          Chr.4: 86268994 - 86292435 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596395_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp1r17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153467.1 NP_703197.1 2 - 3 220 126
          XM_006236545.2 XP_006236607.1 2 - 3 729 126
          XM_008762878.1 XP_008761100.1 3 - 4 656 126
          GenBank mRNA AF294688.1 - 2 - 3 220 126
          FQ213688.1 - 2 - 3 182 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000982.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp1r17

          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of phosphatase activity
          negative regulation of catalytic activity
          protein phosphatase inhibitor activity
          protein serine/threonine phosphatase inhibitor activity
          phosphatase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P379347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109958 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132241 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P379347 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P379347 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P379347 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P379347 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52628 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57263 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10855757 - -
          Affymetrix Rat Gene 1.0 ST Array 10855758 - -
          Affymetrix Rat Gene 1.0 ST Array 10855756 - -
          Affymetrix Rat Gene 1.0 ST Array 10855759 - -
          Affymetrix Rat Gene 1.0 ST Array 10855760 - -
          Affymetrix Rat Gene 1.1 ST Array 10855755 - -
          Rat Genome 230 2.0 Array 1378101_PM_at - -
          Rat Genome 230 2.0 Array 1378101_at - -
          Affymetrix HT Rat Focus Array 1378101_at - -
          Affymetrix HT Rat Focus Array 1378101_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400007055.rn.1 TC0400000982.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400007053.rn.1 TC0400000982.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400007056.rn.1 TC0400000982.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400007059.rn.1 TC0400000982.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400007054.rn.1 TC0400000982.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400007058.rn.1 TC0400000982.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400007057.rn.1 TC0400000982.rn.1 RTA TC
          Rat Expression Set 230 1378101_at - -
          Rat Expression Set 230 1378101_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10855755 - -
          Rat Genome U34 Set rc_AI228404_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21347043 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352190 - -
          RatRef-12 Expression BeadChip ILMN_59848 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0