Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00674109_m1
          See other SYNJ2BP GE Assays ›
          Gene Symbol:
          Synj2bp
          Gene Name:
          synaptojanin 2 binding protein
          Gene Aliases:
          Omp25
          Chromosome Location:
          Chr.6: 105224166 - 105261549 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00674109_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Synj2bp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022599.2 NP_072121.2 2 - 3 281 62
          GenBank mRNA AF260258.1 - 2 - 3 281 62
          BC126067.1 - 2 - 3 269 7323715 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002797.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Synj2bp

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of receptor internalization
          protein targeting
          Rho protein signal transduction
          regulation of endocytosis
          negative regulation of activin receptor signaling pathway
          positive regulation of activin receptor signaling pathway
          intracellular distribution of mitochondria
          positive regulation of protein localization to cell surface
          protein binding
          protein C-terminus binding
          type II activin receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P057261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509439 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139218 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85360 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101413 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3993 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125336 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P057261 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P057261 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P057261 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P729693 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P518958 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P389053 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701897805 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700547385 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700200786 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54075 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15143 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101521 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39888 - -
          Agilent Rat cDNA Microarray (G4105A) 700200786 - -
          Agilent Rat cDNA Microarray (G4105A) 700547385 - -
          Agilent Rat cDNA Microarray (G4105A) 701897805 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10890974 - -
          Affymetrix Rat Gene 1.0 ST Array 10890977 - -
          Affymetrix Rat Gene 1.0 ST Array 10890973 - -
          Affymetrix Rat Gene 1.0 ST Array 10890978 - -
          Affymetrix Rat Gene 1.0 ST Array 10890976 - -
          Affymetrix Rat Gene 1.1 ST Array 10890967 - -
          Rat Genome 230 2.0 Array 1368040_PM_at - -
          Rat Genome 230 2.0 Array 1373585_PM_at - -
          Rat Genome 230 2.0 Array 1368041_at - -
          Rat Genome 230 2.0 Array 1368040_at - -
          Rat Genome 230 2.0 Array 1368039_at - -
          Rat Genome 230 2.0 Array 1368038_at - -
          Rat Genome 230 2.0 Array 1368039_PM_at - -
          Rat Genome 230 2.0 Array 1368038_PM_at - -
          Rat Genome 230 2.0 Array 1373585_at - -
          Rat Genome 230 2.0 Array 1368041_PM_at - -
          Affymetrix HT Rat Focus Array 1368040_PM_at - -
          Affymetrix HT Rat Focus Array 1368039_PM_at - -
          Affymetrix HT Rat Focus Array 1368039_at - -
          Affymetrix HT Rat Focus Array 1368038_PM_at - -
          Affymetrix HT Rat Focus Array 1368040_at - -
          Affymetrix HT Rat Focus Array 1368041_PM_at - -
          Affymetrix HT Rat Focus Array 1368038_at - -
          Affymetrix HT Rat Focus Array 1368041_at - -
          Affymetrix Rat Clariom S Assay PSR0600019717.rn.1 TC0600002797.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600019718.rn.1 TC0600002797.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600019721.rn.1 TC0600002797.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600019720.rn.1 TC0600002797.rn.1 RTA TC
          Rat Expression Set 230 1368039_PM_at - -
          Rat Expression Set 230 1368041_at - -
          Rat Expression Set 230 1368039_at - -
          Rat Expression Set 230 1373585_at - -
          Rat Expression Set 230 1368038_PM_at - -
          Rat Expression Set 230 1373585_PM_at - -
          Rat Expression Set 230 1368040_PM_at - -
          Rat Expression Set 230 1368038_at - -
          Rat Expression Set 230 1368040_at - -
          Rat Expression Set 230 1368041_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10890967 - -
          Rat Genome U34 Set rc_AA964629_at - -
          Rat Genome U34 Set rc_AA924552_at - -
          Rat Genome U34 Set rc_AI172060_at - -
          Rat Genome U34 Set rc_AI070523_at - -
          Rat Genome U34 Set rc_AA859241_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21578590 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360794 - -
          RatRef-12 Expression BeadChip ILMN_49486 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline