Search
Search
| Gene Symbol: Ncdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053543.2 | NP_445995.2 | 2 - 3 | 527 | 66 | |
| XM_006238908.1 | XP_006238970.1 | 2 - 3 | 236 | 66 | ||
| XM_006238909.1 | XP_006238971.1 | 2 - 3 | 297 | 66 | ||
| GenBank mRNA | BC087000.1 | - | 2 - 3 | 527 | 7190905 | 66 |
Set Membership: |
|