Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00686874_m1
          See other LZTS1 GE Assays ›
          Gene Symbol:
          Lzts1
          Gene Name:
          leucine zipper, putative tumor suppressor 1
          Gene Aliases:
          Psdzip70
          Chromosome Location:
          Chr.16: 22250500 - 22306667 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00686874_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lzts1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153470.1 NP_703200.1 2 - 3 693 80
          GenBank mRNA AB075607.1 - 2 - 3 693 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000339.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lzts1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of macroautophagy
          regulation of synaptic plasticity
          regulation of dendrite morphogenesis
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1059903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194355 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26751 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59333 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9776 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165706 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10973 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118725 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1059903 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1059903 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1059903 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1059903 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76208 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6181 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50377 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83048 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10787736 - -
          Affymetrix Rat Gene 1.0 ST Array 10787739 - -
          Affymetrix Rat Gene 1.0 ST Array 10787738 - -
          Affymetrix Rat Gene 1.0 ST Array 10787737 - -
          Affymetrix Rat Gene 1.1 ST Array 10787735 - -
          Rat Genome 230 2.0 Array 1377171_PM_at - -
          Rat Genome 230 2.0 Array 1377171_at - -
          Affymetrix HT Rat Focus Array 1377171_PM_at - -
          Affymetrix HT Rat Focus Array 1377171_at - -
          Affymetrix Rat Clariom S Assay PSR1600003504.rn.1 TC1600000339.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600003505.rn.1 TC1600000339.rn.1 RTA TC
          Rat Expression Set 230 1377171_at - -
          Rat Expression Set 230 1377171_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10787735 - -
          Rat Genome U34 Set rc_AA875041_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21971350 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373484 - -
          RatRef-12 Expression BeadChip ILMN_67553 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline