Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00687271_m1
          See other EAF2 GE Assays ›
          Gene Symbol:
          Eaf2
          Gene Name:
          ELL associated factor 2
          Gene Aliases:
          Traits
          Chromosome Location:
          Chr.11: 66878007 - 66923092 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00687271_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eaf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172047.1 NP_742044.1 5 - 6 869 66
          XM_008768707.1 XP_008766929.1 5 - 6 756 66
          XM_008768708.1 XP_008766930.1 3 - 4 1425 66
          XM_008768709.1 XP_008766931.1 3 - 4 1155 66
          GenBank mRNA AY049022.1 - 5 - 6 869 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000674.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eaf2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          apoptotic process
          biological_process
          negative regulation of cell growth
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of epithelial cell proliferation involved in prostate gland development
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P604033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145571 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140254 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91180 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137413 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P604033 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P604033 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P604033 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P604033 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10512 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12323 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31886 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81793 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41711 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10751402 - -
          Affymetrix Rat Gene 1.0 ST Array 10751398 - -
          Affymetrix Rat Gene 1.0 ST Array 10751397 - -
          Affymetrix Rat Gene 1.0 ST Array 10751399 - -
          Affymetrix Rat Gene 1.0 ST Array 10751400 - -
          Affymetrix Rat Gene 1.0 ST Array 10751401 - -
          Affymetrix Rat Gene 1.0 ST Array 10751403 - -
          Affymetrix Rat Gene 1.1 ST Array 10751396 - -
          Rat Genome 230 2.0 Array 1378247_at - -
          Rat Genome 230 2.0 Array 1378247_PM_at - -
          Affymetrix HT Rat Focus Array 1378247_at - -
          Affymetrix HT Rat Focus Array 1378247_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100003629.rn.1 TC1100000674.rn.1 RTA TC
          Rat Expression Set 230 1378247_PM_at - -
          Rat Expression Set 230 1378247_at - -
          Affymetrix Rat Gene 1.0 ST Array 10751396 - -
          Rat Genome U34 Set rc_AI071961_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22281333 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359394 - -
          RatRef-12 Expression BeadChip ILMN_49312 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline