Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00709992_m1
          See other SCGB1D2 GE Assays ›
          Gene Symbol:
          Scgb1d2
          Gene Name:
          secretoglobin, family 1D, member 2
          Gene Aliases:
          Pbpc1bs, Psbpc1
          Chromosome Location:
          Chr.1: 225591848 - 225600915 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00709992_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scgb1d2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177482.1 NP_803435.1 1 - 2 81 110
          XM_008760227.1 XP_008758449.1 3 - 4 405 110
          XM_008760228.1 XP_008758450.1 3 - 4 536 110
          GenBank mRNA BC099696.1 - 1 - 2 61 110
          V01255.1 - 1 - 2 98 110
          V01545.1 - 1 - 2 81 110

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scgb1d2

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Process(es)

          steroid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P061760 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P007904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38868 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061760 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061760 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P061760 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P482271 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67258 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728590 - -
          Affymetrix Rat Gene 1.0 ST Array 10728591 - -
          Affymetrix Rat Gene 1.0 ST Array 10728592 - -
          Affymetrix Rat Gene 1.1 ST Array 10728589 - -
          Rat Genome 230 2.0 Array 1370858_at - -
          Rat Genome 230 2.0 Array 1370858_PM_at - -
          Affymetrix HT Rat Focus Array 1370858_PM_at - -
          Affymetrix HT Rat Focus Array 1370858_at - -
          Affymetrix Rat Clariom S Assay PSR0100058397.rn.1 TC0100007237.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058395.rn.1 TC0100007237.rn.1 RTA TC
          Rat Expression Set 230 1370858_at - -
          Rat Expression Set 230 1370858_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10728589 - -
          Rat Genome U34 Set J00774cds_s_at - -
          Rat Genome U34 Set V01255_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20918028 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350843 - -
          RatRef-12 Expression BeadChip ILMN_68837 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline