Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710329_m1
          See other MS4A14 GE Assays ›
          Gene Symbol:
          Ms4a14
          Gene Name:
          membrane-spanning 4-domains, subfamily A, member 14
          Gene Aliases:
          Sp3111
          Chromosome Location:
          Chr.1: 227471441 - 227486953 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710329_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ms4a14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181082.2 NP_851596.1 4 - 5 592 115
          GenBank mRNA AY258290.1 - 4 - 5 592 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007282.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ms4a14

          Panther Classification:

          Molecular Function -

          transporter

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P817214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238270 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41085 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155259 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P817214 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P817214 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P817214 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P817214 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29009 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58467 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728903 - -
          Affymetrix Rat Gene 1.0 ST Array 10728898 - -
          Affymetrix Rat Gene 1.0 ST Array 10728900 - -
          Affymetrix Rat Gene 1.0 ST Array 10728899 - -
          Affymetrix Rat Gene 1.0 ST Array 10728901 - -
          Affymetrix Rat Gene 1.0 ST Array 10728902 - -
          Affymetrix Rat Gene 1.1 ST Array 10728897 - -
          Affymetrix Rat Clariom S Assay PSR0100059047.rn.1 TC0100007282.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10728897 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21311797 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650854 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline