Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710391_m1
          See other MAST1 GE Assays ›
          Gene Symbol:
          MAST1
          Gene Name:
          microtubule associated serine/threonine kinase 1
          Gene Aliases:
          Sast
          Chromosome Location:
          Chr.19: 26026045 - 26053762 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710391_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MAST1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181089.1 NP_851603.1 10 - 11 1078 64
          GenBank mRNA AY227206.1 - 10 - 11 1078 64
          AY227207.1 - 10 - 11 1078 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001213.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MAST1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          cytoskeleton organization
          intracellular signal transduction
          magnesium ion binding
          protein serine/threonine kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P132866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336750 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59666 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81117 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P132866 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P132866 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P132866 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16416 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25305 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81216 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806610 - -
          Affymetrix Rat Gene 1.0 ST Array 10806618 - -
          Affymetrix Rat Gene 1.0 ST Array 10806604 - -
          Affymetrix Rat Gene 1.0 ST Array 10806622 - -
          Affymetrix Rat Gene 1.0 ST Array 10806627 - -
          Affymetrix Rat Gene 1.0 ST Array 10806621 - -
          Affymetrix Rat Gene 1.0 ST Array 10806607 - -
          Affymetrix Rat Gene 1.0 ST Array 10806602 - -
          Affymetrix Rat Gene 1.0 ST Array 10806617 - -
          Affymetrix Rat Gene 1.0 ST Array 10806612 - -
          Affymetrix Rat Gene 1.0 ST Array 10806605 - -
          Affymetrix Rat Gene 1.0 ST Array 10806625 - -
          Affymetrix Rat Gene 1.0 ST Array 10806619 - -
          Affymetrix Rat Gene 1.0 ST Array 10806615 - -
          Affymetrix Rat Gene 1.0 ST Array 10806606 - -
          Affymetrix Rat Gene 1.0 ST Array 10806609 - -
          Affymetrix Rat Gene 1.0 ST Array 10806623 - -
          Affymetrix Rat Gene 1.0 ST Array 10806611 - -
          Affymetrix Rat Gene 1.0 ST Array 10806624 - -
          Affymetrix Rat Gene 1.0 ST Array 10806608 - -
          Affymetrix Rat Gene 1.0 ST Array 10806616 - -
          Affymetrix Rat Gene 1.0 ST Array 10806614 - -
          Affymetrix Rat Gene 1.0 ST Array 10806620 - -
          Affymetrix Rat Gene 1.0 ST Array 10806603 - -
          Affymetrix Rat Gene 1.1 ST Array 10806601 - -
          Rat Genome 230 2.0 Array 1391698_at - -
          Rat Genome 230 2.0 Array 1391698_PM_at - -
          Affymetrix HT Rat Focus Array 1391698_at - -
          Affymetrix HT Rat Focus Array 1391698_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900009901.rn.1 TC1900001213.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900009900.rn.1 TC1900001213.rn.1 RTA TC
          Rat Expression Set 230 1391698_at - -
          Rat Expression Set 230 1391698_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806601 - -
          Rat Genome U34 Set rc_AI072348_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21290452 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361933 - -
          RatRef-12 Expression BeadChip ILMN_49986 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline