Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710944_m1
          See other ACSM4 GE Assays ›
          Gene Symbol:
          Acsm4
          Gene Name:
          acyl-CoA synthetase medium-chain family member 4
          Gene Aliases:
          Omacs
          Chromosome Location:
          Chr.1: 189432604 - 189457052 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710944_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acsm4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181695.1 NP_859046.1 3 - 4 532 74
          GenBank mRNA AB096688.1 - 3 - 4 532 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002482.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acsm4

          Panther Classification:

          Molecular Function -

          ligase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid metabolic process
          acyl-CoA metabolic process
          sensory perception of smell
          fatty-acyl-CoA synthase activity
          ATP binding
          odorant binding
          metal ion binding
          butyrate-CoA ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P151358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264533 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10527 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38071 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P151358 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P151358 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P151358 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P269070 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47223 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62773 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66284 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10710291 - -
          Affymetrix Rat Gene 1.0 ST Array 10710290 - -
          Affymetrix Rat Gene 1.0 ST Array 10710287 - -
          Affymetrix Rat Gene 1.0 ST Array 10710282 - -
          Affymetrix Rat Gene 1.0 ST Array 10710283 - -
          Affymetrix Rat Gene 1.0 ST Array 10710281 - -
          Affymetrix Rat Gene 1.0 ST Array 10710293 - -
          Affymetrix Rat Gene 1.0 ST Array 10710288 - -
          Affymetrix Rat Gene 1.0 ST Array 10710292 - -
          Affymetrix Rat Gene 1.0 ST Array 10710289 - -
          Affymetrix Rat Gene 1.0 ST Array 10710284 - -
          Affymetrix Rat Gene 1.0 ST Array 10710294 - -
          Affymetrix Rat Gene 1.0 ST Array 10710285 - -
          Affymetrix Rat Gene 1.0 ST Array 10710286 - -
          Affymetrix Rat Gene 1.1 ST Array 10710280 - -
          Affymetrix Rat Clariom S Assay PSR0100018855.rn.1 TC0100002482.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10710280 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21567193 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368911 - -
          RatRef-12 Expression BeadChip ILMN_64143 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline