Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00788224_m1
          See other CAMK1G GE Assays ›
          Gene Symbol:
          Camk1g
          Gene Name:
          calcium/calmodulin-dependent protein kinase IG
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 112075689 - 112099453 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00788224_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Camk1g
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_182842.1 NP_878262.1 8 - 9 826 71
          XM_006250470.2 XP_006250532.1 8 - 9 943 71
          GenBank mRNA AB101231.1 - 8 - 9 826 71
          AB101232.1 - 8 - 9 826 71
          D86557.1 - 8 - 9 832 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300002248.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Camk1g

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          calcium-mediated signaling
          calmodulin-dependent protein kinase activity
          calmodulin binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P075087 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P097907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241120 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144302 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83072 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P075087 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097907 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P075087 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097907 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P075087 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P097907 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P395522 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15622 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73345 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80304 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770812 - -
          Affymetrix Rat Gene 1.0 ST Array 10770818 - -
          Affymetrix Rat Gene 1.0 ST Array 10770821 - -
          Affymetrix Rat Gene 1.0 ST Array 10770811 - -
          Affymetrix Rat Gene 1.0 ST Array 10770824 - -
          Affymetrix Rat Gene 1.0 ST Array 10770819 - -
          Affymetrix Rat Gene 1.0 ST Array 10770815 - -
          Affymetrix Rat Gene 1.0 ST Array 10770823 - -
          Affymetrix Rat Gene 1.0 ST Array 10770817 - -
          Affymetrix Rat Gene 1.0 ST Array 10770822 - -
          Affymetrix Rat Gene 1.0 ST Array 10770814 - -
          Affymetrix Rat Gene 1.0 ST Array 10770816 - -
          Affymetrix Rat Gene 1.0 ST Array 10770820 - -
          Affymetrix Rat Gene 1.1 ST Array 10770810 - -
          Rat Genome 230 2.0 Array 1391229_at - -
          Rat Genome 230 2.0 Array 1377518_at - -
          Rat Genome 230 2.0 Array 1377518_PM_at - -
          Rat Genome 230 2.0 Array 1391229_PM_at - -
          Affymetrix HT Rat Focus Array 1391229_at - -
          Affymetrix HT Rat Focus Array 1377518_PM_at - -
          Affymetrix HT Rat Focus Array 1391229_PM_at - -
          Affymetrix HT Rat Focus Array 1377518_at - -
          Affymetrix Rat Clariom S Assay PSR1300016051.rn.1 TC1300002248.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016050.rn.1 TC1300002248.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016053.rn.1 TC1300002248.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016056.rn.1 TC1300002248.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016049.rn.1 TC1300002248.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016054.rn.1 TC1300002248.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016052.rn.1 TC1300002248.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016055.rn.1 TC1300002248.rn.1 RTA TC
          Rat Expression Set 230 1377518_PM_at - -
          Rat Expression Set 230 1377518_at - -
          Rat Expression Set 230 1391229_at - -
          Rat Expression Set 230 1391229_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770810 - -
          Rat Genome U34 Set D86557_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20871806 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365119 - -
          RatRef-12 Expression BeadChip ILMN_55982 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline