Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820983_gH
          See other COX6C GE Assays ›
          Gene Symbol:
          Cox6c
          Gene Name:
          cytochrome c oxidase, subunit VIc
          Gene Aliases:
          COVIc, COX-VIc, Cox6c2
          Chromosome Location:
          Chr.7: 74723177 - 74735650 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820983_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cox6c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019360.2 NP_062233.2 3 - 4 330 72
          GenBank mRNA AI716762.1 - 1 - 2 72
          BC058480.1 - 3 - 4 306 6922382 72
          CF976717.1 - 3 - 4 334 72
          FQ210091.1 - 3 - 4 308 72
          FQ210263.1 - 3 - 4 307 72
          FQ217805.1 - 3 - 4 333 72
          FQ221395.1 - 3 - 4 309 72
          FQ222319.1 - 3 - 4 302 72
          FQ222901.1 - 3 - 4 351 72
          FQ223820.1 - 3 - 4 305 72
          K01565.1 - 1 - 2 117 72
          M27466.1 - 3 - 4 341 72
          M27467.1 - 2 - 3 305 72

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cox6c

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          hydrogen ion transmembrane transport
          cytochrome-c oxidase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P146945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581409 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137275 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25697 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P146945 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P146945 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P146945 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P414102 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700368810 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76034 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68186 - -
          Agilent Rat cDNA Microarray (G4105A) 700368810 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10903269 - -
          Affymetrix Rat Gene 1.0 ST Array 10903270 - -
          Affymetrix Rat Gene 1.0 ST Array 10903271 - -
          Affymetrix Rat Gene 1.0 ST Array 10903272 - -
          Affymetrix Rat Gene 1.0 ST Array 10903268 - -
          Affymetrix Rat Gene 1.1 ST Array 10903267 - -
          Rat Genome 230 2.0 Array 1367757_at - -
          Rat Genome 230 2.0 Array 1367757_PM_at - -
          Affymetrix HT Rat Focus Array 1367757_PM_at - -
          Affymetrix HT Rat Focus Array 1367757_at - -
          Affymetrix Rat Clariom S Assay PSR0700023014.rn.1 TC0700002979.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700023015.rn.1 TC0700002979.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700023017.rn.1 TC0700002979.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700023016.rn.1 TC0700002979.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700023018.rn.1 TC0700002979.rn.1 RTA TC
          Rat Expression Set 230 1367757_at - -
          Rat Expression Set 230 1367757_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10903267 - -
          Rat Genome U34 Set M27467_at - -
          Rat Genome U34 Set M27466_at - -
          Rat Genome U34 Set M27466_at - -
          Rat Genome U34 Set M27467_at - -
          Rat Genome U34 Set M27466_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363585 - -
          RatRef-12 Expression BeadChip ILMN_50161 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline