Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821311_g1
          See other TMSB4X GE Assays ›
          Gene Symbol:
          Tmsb4x
          Gene Name:
          thymosin beta 4, X-linked
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 28593405 - 28617267 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821311_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmsb4x
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031136.1 NP_112398.1 2 - 3 168 133
          GenBank mRNA BC058137.1 - 2 - 3 182 6889807 133
          BC084720.1 - 2 - 3 157 133
          BC097308.1 - 2 - 3 167 133
          FQ211846.1 - 2 - 3 175 133
          FQ211950.1 - 2 - 3 178 133
          FQ213262.1 - 2 - 3 175 133
          FQ214330.1 - 2 - 3 175 133
          FQ217532.1 - 2 - 3 175 133
          FQ217617.1 - 2 - 3 175 133
          FQ217622.1 - 2 - 3 175 133
          FQ220985.1 - 2 - 3 175 133
          FQ221085.1 - 2 - 3 175 133
          FQ221124.1 - 2 - 3 175 133
          FQ221299.1 - 2 - 3 175 133
          FQ221364.1 - 2 - 3 175 133
          FQ221505.1 - 2 - 3 175 133
          FQ221785.1 - 2 - 3 175 133
          FQ221964.1 - 2 - 3 175 133
          FQ222011.1 - 2 - 3 175 133
          FQ222205.1 - 2 - 3 175 133
          FQ222334.1 - 2 - 3 175 133
          FQ222360.1 - 2 - 3 169 133
          FQ222463.1 - 2 - 3 175 133
          FQ222519.1 - 2 - 3 175 133
          FQ222755.1 - 2 - 3 175 133
          FQ223266.1 - 2 - 3 172 133
          FQ223325.1 - 2 - 3 175 133
          FQ228401.1 - 2 - 3 175 133
          FQ228412.1 - 2 - 3 175 133
          FQ228477.1 - 2 - 3 175 133
          M26759.1 - 1 - 2 142 133
          M34043.1 - 2 - 3 168 133

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmsb4x

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          actin filament organization
          regulation of cell migration
          sequestering of actin monomers
          actin monomer binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P498066 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P498066 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P498066 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P498066 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P498066 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10734319 - -
          Affymetrix Rat Gene 1.0 ST Array 10876292 - -
          Affymetrix Rat Gene 1.0 ST Array 10876293 - -
          Affymetrix Rat Gene 1.1 ST Array 10734318 - -
          Affymetrix Rat Gene 1.1 ST Array 10876291 - -
          Rat Genome 230 2.0 Array 1387883_a_at - -
          Rat Genome 230 2.0 Array 1387883_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387883_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387883_a_at - -
          Affymetrix Rat Clariom S Assay PSR0X00002145.rn.1 TC0X00000281.rn.1 RTA TC
          Rat Expression Set 230 1387883_a_at - -
          Rat Expression Set 230 1387883_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10734318 - -
          Affymetrix Rat Gene 1.0 ST Array 10876291 - -
          Rat Genome U34 Set M34043_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20854683 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline