Search
Search
| Gene Symbol: Nras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080766.2 | NP_542944.1 | 2 - 3 | 209 | 80 | |
| XM_006233059.2 | XP_006233121.1 | 2 - 3 | 253 | 80 | ||
| GenBank mRNA | BC098659.1 | - | 2 - 3 | 209 | 7368145 | 80 |
Set Membership: |
|