Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00840860_m1
          See other MED9 GE Assays ›
          Gene Symbol:
          Med9
          Gene Name:
          mediator complex subunit 9
          Gene Aliases:
          RGD1563669
          Chromosome Location:
          Chr.10: 46314639 - 46329374 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00840860_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Med9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127302.1 NP_001120774.1 1 - 2 231 67
          GenBank mRNA BC166759.1 - 1 - 2 231 7930828 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000740.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          biological_process
          RNA polymerase II transcription cofactor activity
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P017631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591229 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67782 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017631 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017631 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P017631 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10734035 - -
          Affymetrix Rat Gene 1.0 ST Array 10734034 - -
          Affymetrix Rat Gene 1.1 ST Array 10734033 - -
          Rat Genome 230 2.0 Array 1372924_PM_at - -
          Rat Genome 230 2.0 Array 1372924_at - -
          Affymetrix HT Rat Focus Array 1372924_PM_at - -
          Affymetrix HT Rat Focus Array 1372924_at - -
          Affymetrix Rat Clariom S Assay PSR1000006182.rn.1 TC1000000740.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000006183.rn.1 TC1000000740.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000006180.rn.1 TC1000000740.rn.1 RTA TC
          Rat Expression Set 230 1372924_PM_at - -
          Rat Expression Set 230 1372924_at - -
          Affymetrix Rat Gene 1.0 ST Array 10734033 - -
          Rat Genome U34 Set rc_AA819706_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21751189 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369112 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline