Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01399628_m1
          See other FBXO15 GE Assays ›
          Gene Symbol:
          Fbxo15
          Gene Name:
          F-box protein 15
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 81785408 - 81882796 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01399628_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxo15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108436.1 NP_001101906.1 9 - 10 1324 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000959.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P236548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743875 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36519 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66255 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52179 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171311 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26381 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175390 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66596 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86583 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161401 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5644 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44557 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P236548 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P236548 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P236548 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P236548 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50569 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65160 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99929 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55573 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87616 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10803029 - -
          Affymetrix Rat Gene 1.0 ST Array 10803034 - -
          Affymetrix Rat Gene 1.0 ST Array 10803027 - -
          Affymetrix Rat Gene 1.0 ST Array 10803032 - -
          Affymetrix Rat Gene 1.0 ST Array 10803033 - -
          Affymetrix Rat Gene 1.0 ST Array 10803031 - -
          Affymetrix Rat Gene 1.0 ST Array 10803028 - -
          Affymetrix Rat Gene 1.0 ST Array 10803030 - -
          Affymetrix Rat Gene 1.1 ST Array 10803025 - -
          Rat Genome 230 2.0 Array 1380523_at - -
          Rat Genome 230 2.0 Array 1380523_PM_at - -
          Affymetrix HT Rat Focus Array 1380523_at - -
          Affymetrix HT Rat Focus Array 1380523_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800006796.rn.1 TC1800000959.rn.1 RTA TC
          Rat Expression Set 230 1380523_at - -
          Rat Expression Set 230 1380523_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10803025 - -
          Rat Genome U34 Set rc_AI145917_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22382929 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355981 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline