Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01400706_m1
          See other NHEJ1 GE Assays ›
          Gene Symbol:
          Nhej1
          Gene Name:
          nonhomologous end-joining factor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 82230230 - 82327923 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01400706_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nhej1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014217.1 NP_001014239.1 5 - 6 709 92
          XM_006245240.1 XP_006245302.1 5 - 6 787 92
          XM_006245241.2 XP_006245303.1 5 - 6 1363 92
          XM_006245242.2 XP_006245304.1 5 - 6 1132 92
          GenBank mRNA BC079033.1 - 5 - 6 709 7114269 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002257.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nhej1

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via nonhomologous end joining
          DNA recombination
          biological_process
          response to ionizing radiation
          B cell differentiation
          T cell differentiation
          molecular_function
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P760697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168325 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93649 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106344 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66004 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166166 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18729 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117757 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133476 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16222 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154525 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P760697 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P760697 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P760697 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P387324 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P760697 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98646 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102497 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72457 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70973 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28656 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90302 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61802 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10929033 - -
          Affymetrix Rat Gene 1.0 ST Array 10929038 - -
          Affymetrix Rat Gene 1.0 ST Array 10929035 - -
          Affymetrix Rat Gene 1.0 ST Array 10929039 - -
          Affymetrix Rat Gene 1.0 ST Array 10929036 - -
          Affymetrix Rat Gene 1.0 ST Array 10929034 - -
          Affymetrix Rat Gene 1.0 ST Array 10929032 - -
          Affymetrix Rat Gene 1.0 ST Array 10929040 - -
          Affymetrix Rat Gene 1.0 ST Array 10929037 - -
          Affymetrix Rat Gene 1.1 ST Array 10929031 - -
          Rat Genome 230 2.0 Array 1374245_at - -
          Rat Genome 230 2.0 Array 1374245_PM_at - -
          Affymetrix HT Rat Focus Array 1374245_PM_at - -
          Affymetrix HT Rat Focus Array 1374245_at - -
          Affymetrix Rat Clariom S Assay PSR0900018382.rn.1 TC0900002257.rn.1 RTA TC
          Rat Expression Set 230 1374245_PM_at - -
          Rat Expression Set 230 1374245_at - -
          Affymetrix Rat Gene 1.0 ST Array 10929031 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355080 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline