Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01401528_m1
          See other RPE GE Assays ›
          Gene Symbol:
          Rpe
          Gene Name:
          ribulose-5-phosphate-3-epimerase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 73687682 - 73713643 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01401528_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpe
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033903.1 NP_001029075.1 1 - 2 131 64
          XM_006245104.2 XP_006245166.1 1 - 2 227 64
          XM_006245105.2 XP_006245167.1 1 - 2 230 64
          XM_006245106.2 XP_006245168.1 1 - 2 226 64
          XM_006245107.2 XP_006245169.1 1 - 2 227 64
          XM_006245109.2 XP_006245171.1 1 - 2 223 64
          XM_008767287.1 XP_008765509.1 1 - 2 231 64
          XM_008767288.1 XP_008765510.1 1 - 2 235 64
          GenBank mRNA BC101869.1 - 1 - 2 131 5621971 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000821.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpe

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          pentose-phosphate shunt
          pentose-phosphate shunt, non-oxidative branch
          pentose catabolic process
          cellular carbohydrate metabolic process
          ribulose-phosphate 3-epimerase activity
          carbohydrate binding
          identical protein binding
          protein homodimerization activity
          metal ion binding
          monosaccharide binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P652890 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P248663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299536 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119294 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177696 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P652890 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P248663 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P652890 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P248663 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P652890 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P248663 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P248663 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P652890 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42747 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4625 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10924077 - -
          Affymetrix Rat Gene 1.0 ST Array 10924079 - -
          Affymetrix Rat Gene 1.0 ST Array 10924080 - -
          Affymetrix Rat Gene 1.0 ST Array 10924078 - -
          Affymetrix Rat Gene 1.0 ST Array 10924083 - -
          Affymetrix Rat Gene 1.0 ST Array 10924082 - -
          Affymetrix Rat Gene 1.1 ST Array 10924076 - -
          Rat Genome 230 2.0 Array 1389202_at - -
          Rat Genome 230 2.0 Array 1389202_PM_at - -
          Affymetrix HT Rat Focus Array 1389202_at - -
          Affymetrix HT Rat Focus Array 1389202_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900006815.rn.1 TC0900000821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900006813.rn.1 TC0900000821.rn.1 RTA TC
          Rat Expression Set 230 1389202_at - -
          Rat Expression Set 230 1389202_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10924076 - -
          Rat Genome U34 Set rc_AA944520_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22319811 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline