Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01404249_m1
          See other TOP3B GE Assays ›
          Gene Symbol:
          Top3b
          Gene Name:
          topoisomerase (DNA) III beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 88346305 - 88374897 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01404249_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Top3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105861.1 NP_001099331.1 18 - 19 2379 57
          XM_006248671.2 XP_006248733.1 17 - 18 2513 57
          XM_006248673.2 XP_006248735.1 17 - 18 2325 57
          GenBank mRNA FQ211004.1 - 17 - 18 2326 57
          FQ212059.1 - 16 - 17 3053 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100002027.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA topological change
          chromosome segregation
          DNA binding
          DNA topoisomerase type I activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P141564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653657 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130375 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1998 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44736 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156728 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141564 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141564 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P141564 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P716940 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32643 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45090 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10755835 - -
          Affymetrix Rat Gene 1.0 ST Array 10755837 - -
          Affymetrix Rat Gene 1.0 ST Array 10755825 - -
          Affymetrix Rat Gene 1.0 ST Array 10755826 - -
          Affymetrix Rat Gene 1.0 ST Array 10755829 - -
          Affymetrix Rat Gene 1.0 ST Array 10755828 - -
          Affymetrix Rat Gene 1.0 ST Array 10755836 - -
          Affymetrix Rat Gene 1.0 ST Array 10755824 - -
          Affymetrix Rat Gene 1.0 ST Array 10755834 - -
          Affymetrix Rat Gene 1.0 ST Array 10755830 - -
          Affymetrix Rat Gene 1.0 ST Array 10755838 - -
          Affymetrix Rat Gene 1.0 ST Array 10755827 - -
          Affymetrix Rat Gene 1.0 ST Array 10755839 - -
          Affymetrix Rat Gene 1.0 ST Array 10755831 - -
          Affymetrix Rat Gene 1.0 ST Array 10755823 - -
          Affymetrix Rat Gene 1.0 ST Array 10755832 - -
          Affymetrix Rat Gene 1.0 ST Array 10755833 - -
          Affymetrix Rat Gene 1.1 ST Array 10755822 - -
          Rat Genome 230 2.0 Array 1373553_PM_at - -
          Rat Genome 230 2.0 Array 1373553_at - -
          Affymetrix HT Rat Focus Array 1373553_at - -
          Affymetrix HT Rat Focus Array 1373553_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100013419.rn.1 TC1100002027.rn.1 RTA TC
          Rat Expression Set 230 1373553_at - -
          Rat Expression Set 230 1373553_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10755822 - -
          Rat Genome U34 Set rc_AI233765_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21348821 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363550 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline