Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01405825_m1
          See other NRG2 GE Assays ›
          Gene Symbol:
          Nrg2
          Gene Name:
          neuregulin 2
          Gene Aliases:
          NTAK_alpha2a, Ntak
          Chromosome Location:
          Chr.18: 28835391 - 29015552 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01405825_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nrg2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001136151.1 NP_001129623.1 6 - 7 1574 60
          GenBank mRNA AB001576.1 - 6 - 7 685 60
          D89995.1 - 7 - 8 1444 60
          D89996.1 - 6 - 7 1574 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001262.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          epidermal growth factor receptor signaling pathway
          intracellular signal transduction
          animal organ development
          epidermal growth factor-activated receptor activity
          receptor binding
          epidermal growth factor receptor binding
          growth factor activity
          ErbB-3 class receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P053115 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P053114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133547 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18318 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26245 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178768 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126016 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123943 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125622 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63472 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97680 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25797 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177217 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160509 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162826 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144625 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15148 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179538 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51721 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54124 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126792 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92137 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50036 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139568 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72058 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79012 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P053114 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P053115 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P053114 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P053115 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P053114 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P053115 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P372485 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P452588 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P311455 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16834 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78637 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36800 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26084 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46941 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96923 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86914 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33749 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16566 - -
          Agilent Rat cDNA Microarray (G4105A) 702451955 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10803937 - -
          Affymetrix Rat Gene 1.0 ST Array 10803930 - -
          Affymetrix Rat Gene 1.0 ST Array 10803933 - -
          Affymetrix Rat Gene 1.0 ST Array 10803941 - -
          Affymetrix Rat Gene 1.0 ST Array 10803939 - -
          Affymetrix Rat Gene 1.0 ST Array 10803942 - -
          Affymetrix Rat Gene 1.0 ST Array 10803940 - -
          Affymetrix Rat Gene 1.0 ST Array 10803935 - -
          Affymetrix Rat Gene 1.0 ST Array 10803938 - -
          Affymetrix Rat Gene 1.0 ST Array 10803931 - -
          Affymetrix Rat Gene 1.0 ST Array 10803934 - -
          Affymetrix Rat Gene 1.0 ST Array 10803932 - -
          Affymetrix Rat Gene 1.1 ST Array 10803929 - -
          Rat Genome 230 2.0 Array 1387990_at - -
          Rat Genome 230 2.0 Array 1387990_PM_at - -
          Rat Genome 230 2.0 Array 1391033_PM_s_at - -
          Rat Genome 230 2.0 Array 1391033_s_at - -
          Rat Genome 230 2.0 Array 1390044_PM_at - -
          Rat Genome 230 2.0 Array 1390044_at - -
          Affymetrix HT Rat Focus Array 1390044_at - -
          Affymetrix HT Rat Focus Array 1391033_s_at - -
          Affymetrix HT Rat Focus Array 1387990_PM_at - -
          Affymetrix HT Rat Focus Array 1390044_PM_at - -
          Affymetrix HT Rat Focus Array 1391033_PM_s_at - -
          Affymetrix HT Rat Focus Array 1387990_at - -
          Affymetrix Rat Clariom S Assay PSR1800008997.rn.1 TC1800001262.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800008998.rn.1 TC1800001262.rn.1 RTA TC
          Rat Expression Set 230 1390044_PM_at - -
          Rat Expression Set 230 1387990_PM_at - -
          Rat Expression Set 230 1390044_at - -
          Rat Expression Set 230 1387990_at - -
          Rat Expression Set 230 1391033_PM_s_at - -
          Rat Expression Set 230 1391033_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10803929 - -
          Rat Genome U34 Set AB001576_s_at - -
          Rat Genome U34 Set rc_AI102053_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21137692 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350614 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline