Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01406468_g1
          See other YIPF5 GE Assays ›
          Gene Symbol:
          Yipf5
          Gene Name:
          Yip1 domain family, member 5
          Gene Aliases:
          Ac2-256
          Chromosome Location:
          Chr.18: 33401103 - 33436885 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01406468_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Yipf5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014150.3 NP_001014172.3 4 - 5 533 78
          XM_006254664.2 XP_006254726.1 13 - 14 1694 78
          GenBank mRNA AY321347.1 - 13 - 14 1700 78
          BC083754.1 - 4 - 5 533 78

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Yipf5

          Gene Ontology Categories:

          Function(s)

          protein transport
          vesicle-mediated transport
          regulation of ER to Golgi vesicle-mediated transport

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P219759 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P161724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62520 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102984 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P219759 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P161724 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P219759 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P161724 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P219759 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P161724 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P219759 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P260773 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38350 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51031 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100159 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2468 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10804148 - -
          Affymetrix Rat Gene 1.0 ST Array 10804146 - -
          Affymetrix Rat Gene 1.0 ST Array 10804151 - -
          Affymetrix Rat Gene 1.1 ST Array 10804145 - -
          Rat Genome 230 2.0 Array 1371843_PM_at - -
          Rat Genome 230 2.0 Array 1371843_at - -
          Affymetrix HT Rat Focus Array 1371843_PM_at - -
          Affymetrix HT Rat Focus Array 1371843_at - -
          Affymetrix Rat Clariom S Assay PSR1800009523.rn.1 TC1800001327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800009522.rn.1 TC1800001327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800009521.rn.1 TC1800001327.rn.1 RTA TC
          Rat Expression Set 230 1371843_at - -
          Rat Expression Set 230 1371843_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10804145 - -
          Rat Genome U34 Set rc_AI234128_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650077 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline