Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01407207_m1
          See other IGSF5 GE Assays ›
          Gene Symbol:
          Igsf5
          Gene Name:
          immunoglobulin superfamily, member 5
          Gene Aliases:
          Jam4
          Chromosome Location:
          Chr.11: 36736579 - 36779614 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01407207_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igsf5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001006990.2 NP_001006991.1 8 - 9 1376 81
          XM_006248155.2 XP_006248217.1 8 - 9 1241 81
          XM_006248156.2 XP_006248218.1 7 - 8 940 81
          GenBank mRNA AY439284.1 - 7 - 8 1022 81
          BC086342.1 - 8 - 9 1082 81
          BC097947.1 - 8 - 9 1376 7373302 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000358.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Igsf5

          Panther Classification:

          Molecular Function -

          defense/immunity protein immunoglobulin

          Gene Ontology Categories:

          Function(s) Process(es)

          single organismal cell-cell adhesion
          regulation of neurogenesis
          PDZ domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P161691 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P18730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503041 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67724 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8335 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132882 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47971 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122562 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93122 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18730 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P161691 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P161691 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18730 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P18730 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P161691 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P250186 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18730 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12433 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1507 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37482 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104570 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750500 - -
          Affymetrix Rat Gene 1.0 ST Array 10750498 - -
          Affymetrix Rat Gene 1.0 ST Array 10750495 - -
          Affymetrix Rat Gene 1.0 ST Array 10750504 - -
          Affymetrix Rat Gene 1.0 ST Array 10750496 - -
          Affymetrix Rat Gene 1.0 ST Array 10750497 - -
          Affymetrix Rat Gene 1.0 ST Array 10750503 - -
          Affymetrix Rat Gene 1.0 ST Array 10750492 - -
          Affymetrix Rat Gene 1.0 ST Array 10750499 - -
          Affymetrix Rat Gene 1.0 ST Array 10753373 - -
          Affymetrix Rat Gene 1.0 ST Array 10750493 - -
          Affymetrix Rat Gene 1.1 ST Array 10750491 - -
          Affymetrix Rat Gene 1.1 ST Array 10753372 - -
          Rat Genome 230 2.0 Array 1385767_PM_at - -
          Rat Genome 230 2.0 Array 1385767_at - -
          Affymetrix HT Rat Focus Array 1385767_at - -
          Affymetrix HT Rat Focus Array 1385767_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100001768.rn.1 TC1100000358.rn.1 RTA TC
          Rat Expression Set 230 1385767_at - -
          Rat Expression Set 230 1385767_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10750491 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22260948 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376918 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline