Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01408186_m1
          See other SPPL2B GE Assays ›
          Gene Symbol:
          Sppl2b
          Gene Name:
          signal peptide peptidase-like 2B
          Gene Aliases:
          RGD1308556
          Chromosome Location:
          Chr.7: 11712187 - 11724911 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01408186_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sppl2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014200.1 NP_001014222.1 8 - 9 945 127
          XM_006240947.2 XP_006241009.1 8 - 9 1015 127
          XM_006240948.2 XP_006241010.1 8 - 9 1012 127
          XM_006240952.2 XP_006241014.1 8 - 9 1015 127
          XM_008765108.1 XP_008763330.1 7 - 8 1453 127
          XM_008765109.1 XP_008763331.1 7 - 8 1450 127
          XM_008765110.1 XP_008763332.1 7 - 8 1453 127
          XM_008765111.1 XP_008763333.1 8 - 9 1016 127
          XM_008765112.1 XP_008763334.1 8 - 9 1013 127
          XM_008765113.1 XP_008763335.1 7 - 8 1453 127
          XM_008765114.1 XP_008763336.1 7 - 8 1453 127
          XM_008765115.1 XP_008763337.1 7 - 8 1450 127
          XM_008765116.1 XP_008763338.1 7 - 8 1453 127
          XM_008765117.1 XP_008763339.1 7 - 8 1453 127
          GenBank mRNA BC087132.1 - 8 - 9 945 7121303 127
          FQ223857.1 - 9 - 10 1095 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002133.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sppl2b

          Panther Classification:

          Molecular Function -

          aspartic protease

          Gene Ontology Categories:

          Function(s) Process(es)

          membrane protein ectodomain proteolysis
          membrane protein intracellular domain proteolysis
          regulation of immune response
          aspartic endopeptidase activity, intramembrane cleaving
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P049027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67790 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57062 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10159 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049027 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049027 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P049027 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P539244 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82498 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63413 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10900391 - -
          Affymetrix Rat Gene 1.0 ST Array 10900394 - -
          Affymetrix Rat Gene 1.0 ST Array 10900388 - -
          Affymetrix Rat Gene 1.0 ST Array 10900395 - -
          Affymetrix Rat Gene 1.0 ST Array 10900387 - -
          Affymetrix Rat Gene 1.0 ST Array 10900397 - -
          Affymetrix Rat Gene 1.0 ST Array 10900393 - -
          Affymetrix Rat Gene 1.0 ST Array 10900384 - -
          Affymetrix Rat Gene 1.0 ST Array 10900396 - -
          Affymetrix Rat Gene 1.0 ST Array 10900390 - -
          Affymetrix Rat Gene 1.0 ST Array 10900385 - -
          Affymetrix Rat Gene 1.0 ST Array 10900389 - -
          Affymetrix Rat Gene 1.0 ST Array 10900392 - -
          Affymetrix Rat Gene 1.0 ST Array 10900398 - -
          Affymetrix Rat Gene 1.0 ST Array 10900386 - -
          Affymetrix Rat Gene 1.1 ST Array 10900383 - -
          Rat Genome 230 2.0 Array 1374229_PM_at - -
          Rat Genome 230 2.0 Array 1374229_at - -
          Affymetrix HT Rat Focus Array 1374229_at - -
          Affymetrix HT Rat Focus Array 1374229_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700016767.rn.1 TC0700002133.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700016769.rn.1 TC0700002133.rn.1 RTA TC
          Rat Expression Set 230 1374229_PM_at - -
          Rat Expression Set 230 1374229_at - -
          Affymetrix Rat Gene 1.0 ST Array 10900383 - -
          Rat Genome U34 Set rc_AI030564_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21229230 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline