Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01409547_m1
          See other RSPH10B GE Assays ›
          Gene Symbol:
          Rsph10b
          Gene Name:
          radial spoke head 10 homolog B (Chlamydomonas)
          Gene Aliases:
          RGD1311893
          Chromosome Location:
          Chr.12: 12665194 - 12713347 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01409547_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rsph10b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013867.1 NP_001013889.1 18 - 19 2410 102
          XM_006248858.2 XP_006248920.1 19 - 20 3237 102
          XM_006248859.2 XP_006248921.1 19 - 20 3105 102
          XM_006248860.2 XP_006248922.1 19 - 20 3225 102
          XM_006248861.2 XP_006248923.1 18 - 19 3146 102
          GenBank mRNA BC081935.1 - 18 - 19 2410 7114952 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001107.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rsph10b

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P19822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799746 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25791 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120607 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65209 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45992 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19822 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19822 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P19822 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P506092 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47795 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20842 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94530 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10760334 - -
          Affymetrix Rat Gene 1.0 ST Array 10760335 - -
          Affymetrix Rat Gene 1.0 ST Array 10760338 - -
          Affymetrix Rat Gene 1.0 ST Array 10760332 - -
          Affymetrix Rat Gene 1.0 ST Array 10760323 - -
          Affymetrix Rat Gene 1.0 ST Array 10760341 - -
          Affymetrix Rat Gene 1.0 ST Array 10760328 - -
          Affymetrix Rat Gene 1.0 ST Array 10760331 - -
          Affymetrix Rat Gene 1.0 ST Array 10760329 - -
          Affymetrix Rat Gene 1.0 ST Array 10760325 - -
          Affymetrix Rat Gene 1.0 ST Array 10760337 - -
          Affymetrix Rat Gene 1.0 ST Array 10760342 - -
          Affymetrix Rat Gene 1.0 ST Array 10760330 - -
          Affymetrix Rat Gene 1.0 ST Array 10760326 - -
          Affymetrix Rat Gene 1.0 ST Array 10760339 - -
          Affymetrix Rat Gene 1.0 ST Array 10760333 - -
          Affymetrix Rat Gene 1.0 ST Array 10760340 - -
          Affymetrix Rat Gene 1.0 ST Array 10760324 - -
          Affymetrix Rat Gene 1.0 ST Array 10760327 - -
          Affymetrix Rat Gene 1.1 ST Array 10760322 - -
          Rat Genome 230 2.0 Array 1381340_at - -
          Rat Genome 230 2.0 Array 1381340_PM_at - -
          Affymetrix HT Rat Focus Array 1381340_PM_at - -
          Affymetrix HT Rat Focus Array 1381340_at - -
          Affymetrix Rat Clariom S Assay PSR1200009454.rn.1 TC1200001107.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200009455.rn.1 TC1200001107.rn.1 RTA TC
          Rat Expression Set 230 1381340_PM_at - -
          Rat Expression Set 230 1381340_at - -
          Affymetrix Rat Gene 1.0 ST Array 10760322 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21441576 - -
          ABI Rat Genome Survey Microarray 22121466 - -
          ABI Rat Genome Survey Microarray 22406918 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352202 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0