Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01410403_g1
          See other DNAJB6 GE Assays ›
          Gene Symbol:
          Dnajb6
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily B, member 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 2711339 - 2774969 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01410403_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnajb6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013209.1 NP_001013227.1 6 - 7 593 79
          XM_006235863.2 XP_006235925.1 6 - 7 639 79
          XM_006235864.2 XP_006235926.1 7 - 8 696 79
          XM_006235866.2 XP_006235928.1 6 - 7 636 79
          GenBank mRNA BC078908.1 - 6 - 7 593 7113862 79

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnajb6

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          actin cytoskeleton organization
          extracellular matrix organization
          positive regulation of ATPase activity
          protein localization to nucleus
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          intermediate filament organization
          negative regulation of transcription, DNA-templated
          chorio-allantoic fusion
          syncytiotrophoblast cell differentiation involved in labyrinthine layer development
          chorion development
          negative regulation of inclusion body assembly
          ATPase activator activity
          DNA binding
          heat shock protein binding
          unfolded protein binding
          chaperone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P024856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951274 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65044 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156469 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175678 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141897 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8365 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44860 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40498 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P024856 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P024856 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P024856 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P386840 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P840778 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44614 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74284 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76906 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70045 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24560 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728465 - -
          Affymetrix Rat Gene 1.0 ST Array 10840641 - -
          Affymetrix Rat Gene 1.0 ST Array 10850604 - -
          Affymetrix Rat Gene 1.0 ST Array 10840649 - -
          Affymetrix Rat Gene 1.0 ST Array 10850606 - -
          Affymetrix Rat Gene 1.0 ST Array 10859809 - -
          Affymetrix Rat Gene 1.0 ST Array 10859813 - -
          Affymetrix Rat Gene 1.0 ST Array 10859808 - -
          Affymetrix Rat Gene 1.0 ST Array 10850605 - -
          Affymetrix Rat Gene 1.0 ST Array 10859812 - -
          Affymetrix Rat Gene 1.0 ST Array 10850607 - -
          Affymetrix Rat Gene 1.1 ST Array 10840640 - -
          Affymetrix Rat Gene 1.1 ST Array 10850603 - -
          Affymetrix Rat Gene 1.1 ST Array 10728464 - -
          Affymetrix Rat Gene 1.1 ST Array 10859807 - -
          Rat Genome 230 2.0 Array 1382024_at - -
          Rat Genome 230 2.0 Array 1382024_PM_at - -
          Rat Genome 230 2.0 Array 1391422_at - -
          Rat Genome 230 2.0 Array 1391422_PM_at - -
          Affymetrix HT Rat Focus Array 1382024_at - -
          Affymetrix HT Rat Focus Array 1382024_PM_at - -
          Affymetrix HT Rat Focus Array 1391422_PM_at - -
          Affymetrix HT Rat Focus Array 1391422_at - -
          Affymetrix Rat Clariom S Assay PSR0400000310.rn.1 TC0400000055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400000305.rn.1 TC0400000055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400000306.rn.1 TC0400000055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400000313.rn.1 TC0400000055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400000307.rn.1 TC0400000055.rn.1 RTA TC
          Rat Expression Set 230 1382024_at - -
          Rat Expression Set 230 1391422_at - -
          Rat Expression Set 230 1382024_PM_at - -
          Rat Expression Set 230 1391422_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10859807 - -
          Rat Genome U34 Set rc_AI231687_at - -
          Rat Genome U34 Set rc_AI179560_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21722959 - -
          ABI Rat Genome Survey Microarray 21748549 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370974 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline