Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01411350_m1
          See other HOGA1 GE Assays ›
          Gene Symbol:
          Hoga1
          Gene Name:
          4-hydroxy-2-oxoglutarate aldolase 1
          Gene Aliases:
          Dhdpsl, Npl2, RGD1310475
          Chromosome Location:
          Chr.1: 261291743 - 261319742 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01411350_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hoga1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106355.1 NP_001099825.1 6 - 7 1037 88
          XM_006231386.2 XP_006231448.1 5 - 6 1035 88
          GenBank mRNA FQ210513.1 - 6 - 7 851 88
          FQ218423.1 - 6 - 7 849 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003629.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          lyase

          Gene Ontology Categories:

          Function(s) Process(es)

          glyoxylate catabolic process
          4-hydroxyproline catabolic process
          oxalate metabolic process
          pyruvate biosynthetic process
          glyoxylate metabolic process
          4-hydroxy-2-oxoglutarate aldolase activity
          lyase activity
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P088575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808027 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152516 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57496 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62965 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088575 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088575 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P088575 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P555417 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68603 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63417 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11114 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10730324 - -
          Affymetrix Rat Gene 1.0 ST Array 10730326 - -
          Affymetrix Rat Gene 1.0 ST Array 10730325 - -
          Affymetrix Rat Gene 1.0 ST Array 10730328 - -
          Affymetrix Rat Gene 1.0 ST Array 10730329 - -
          Affymetrix Rat Gene 1.0 ST Array 10730327 - -
          Affymetrix Rat Gene 1.0 ST Array 10730323 - -
          Affymetrix Rat Gene 1.1 ST Array 10730322 - -
          Rat Genome 230 2.0 Array 1375560_PM_at - -
          Rat Genome 230 2.0 Array 1389839_at - -
          Rat Genome 230 2.0 Array 1375560_at - -
          Rat Genome 230 2.0 Array 1389839_PM_at - -
          Rat Genome 230 2.0 Array 1390421_at - -
          Rat Genome 230 2.0 Array 1390421_PM_at - -
          Affymetrix HT Rat Focus Array 1389839_PM_at - -
          Affymetrix HT Rat Focus Array 1375560_PM_at - -
          Affymetrix HT Rat Focus Array 1389839_at - -
          Affymetrix HT Rat Focus Array 1375560_at - -
          Affymetrix Rat Clariom S Assay PSR0100029556.rn.1 TC0100003629.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029554.rn.1 TC0100003629.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029557.rn.1 TC0100003629.rn.1 RTA TC
          Rat Expression Set 230 1375560_PM_at - -
          Rat Expression Set 230 1375560_at - -
          Rat Expression Set 230 1389839_PM_at - -
          Rat Expression Set 230 1389839_at - -
          Rat Expression Set 230 1390421_at - -
          Rat Expression Set 230 1390421_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10730322 - -
          Rat Genome U34 Set rc_AA892488_at - -
          Rat Genome U34 Set rc_AA892488_g_at - -
          Rat Genome U34 Set rc_AA945579_at - -
          Rat Genome U34 Set rc_AI113256_s_at - -
          Rat Genome U34 Set rc_AI232524_at - -
          Rat Genome U34 Set rc_AI233647_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21838101 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366025 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline