Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01413122_m1
          See other LRRC66 GE Assays ›
          Gene Symbol:
          Lrrc66
          Gene Name:
          leucine rich repeat containing 66
          Gene Aliases:
          RGD1306094
          Chromosome Location:
          Chr.14: 37128698 - 37153890 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01413122_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc66
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008763.1 NP_001008763.1 3 - 4 828 98
          XM_008770138.1 XP_008768360.1 4 - 5 1830 98
          GenBank mRNA AY383699.1 - 3 - 4 828 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000438.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lrrc66

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P048446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681918 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107249 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33004 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P048446 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P048446 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P048446 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P208102 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62590 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66845 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10772328 - -
          Affymetrix Rat Gene 1.0 ST Array 10772326 - -
          Affymetrix Rat Gene 1.0 ST Array 10772329 - -
          Affymetrix Rat Gene 1.0 ST Array 10772327 - -
          Affymetrix Rat Gene 1.1 ST Array 10772325 - -
          Affymetrix Rat Clariom S Assay PSR1400003384.rn.1 TC1400000438.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10772325 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21914504 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376362 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline