Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01414006_g1
          See other RCHY1 GE Assays ›
          Gene Symbol:
          Rchy1
          Gene Name:
          ring finger and CHY zinc finger domain containing 1, E3 ubiquitin protein ligase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 17615968 - 17631715 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01414006_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rchy1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007618.1 NP_001007619.1 8 - 9 1043 82
          GenBank mRNA BC083739.1 - 8 - 9 1043 7191744 82
          FQ216834.1 - 8 - 9 810 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Androgen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rchy1

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein ubiquitination
          positive regulation of protein ubiquitination
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          protein autoubiquitination
          p53 binding
          ubiquitin-protein transferase activity
          receptor binding
          zinc ion binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1025750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649477 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152959 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93216 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1025750 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1025750 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1025750 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1025750 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30854 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32975 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10771752 - -
          Affymetrix Rat Gene 1.0 ST Array 10771755 - -
          Affymetrix Rat Gene 1.0 ST Array 10771754 - -
          Affymetrix Rat Gene 1.0 ST Array 10771753 - -
          Affymetrix Rat Gene 1.0 ST Array 10771756 - -
          Affymetrix Rat Gene 1.0 ST Array 10771757 - -
          Affymetrix Rat Gene 1.0 ST Array 10771759 - -
          Affymetrix Rat Gene 1.0 ST Array 10771758 - -
          Affymetrix Rat Gene 1.1 ST Array 10771751 - -
          Rat Genome 230 2.0 Array 1388651_at - -
          Rat Genome 230 2.0 Array 1388651_PM_at - -
          Affymetrix HT Rat Focus Array 1388651_PM_at - -
          Affymetrix HT Rat Focus Array 1388651_at - -
          Affymetrix Rat Clariom S Assay PSR1400002127.rn.1 TC1400000263.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400002128.rn.1 TC1400000263.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400002129.rn.1 TC1400000263.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400002126.rn.1 TC1400000263.rn.1 RTA TC
          Rat Expression Set 230 1388651_at - -
          Rat Expression Set 230 1388651_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10771751 - -
          Rat Genome U34 Set rc_AA800053_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21679487 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356257 - -
          RatRef-12 Expression BeadChip ILMN_65668 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline