Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01414261_m1
          See other RASSF6 GE Assays ›
          Gene Symbol:
          Rassf6
          Gene Name:
          Ras association (RalGDS/AF-6) domain family member 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 18983855 - 19023921 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01414261_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rassf6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025671.1 NP_001020842.1 7 - 8 783 82
          XM_006250745.2 XP_006250807.1 8 - 9 895 82
          XM_006250747.2 XP_006250809.1 8 - 9 883 82
          XM_006250748.1 XP_006250810.1 8 - 9 918 82
          XM_008770036.1 XP_008768258.1 8 - 9 978 82
          XM_008770037.1 XP_008768259.1 8 - 9 928 82
          XM_008770038.1 XP_008768260.1 8 - 9 4170 82
          GenBank mRNA BC097375.1 - 7 - 8 783 7444629 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000279.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rassf6

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s)

          apoptotic process
          signal transduction
          regulation of apoptotic process
          positive regulation of apoptotic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P436040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406502 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118747 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32490 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170396 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P436040 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P436040 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P436040 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P436040 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29192 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86266 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30904 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19026 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10771776 - -
          Affymetrix Rat Gene 1.0 ST Array 10771773 - -
          Affymetrix Rat Gene 1.0 ST Array 10771780 - -
          Affymetrix Rat Gene 1.0 ST Array 10771781 - -
          Affymetrix Rat Gene 1.0 ST Array 10771775 - -
          Affymetrix Rat Gene 1.0 ST Array 10771782 - -
          Affymetrix Rat Gene 1.0 ST Array 10771774 - -
          Affymetrix Rat Gene 1.0 ST Array 10771778 - -
          Affymetrix Rat Gene 1.0 ST Array 10771777 - -
          Affymetrix Rat Gene 1.0 ST Array 10771779 - -
          Affymetrix Rat Gene 1.1 ST Array 10771771 - -
          Rat Genome 230 2.0 Array 1382384_at - -
          Rat Genome 230 2.0 Array 1382384_PM_at - -
          Affymetrix HT Rat Focus Array 1382384_PM_at - -
          Affymetrix HT Rat Focus Array 1382384_at - -
          Affymetrix Rat Clariom S Assay PSR1400002207.rn.1 TC1400000279.rn.1 RTA TC
          Rat Expression Set 230 1382384_PM_at - -
          Rat Expression Set 230 1382384_at - -
          Affymetrix Rat Gene 1.0 ST Array 10771771 - -
          Rat Genome U34 Set rc_AI029149_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21632382 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354385 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline