Search
Search
| Gene Symbol: Psmc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100509.1 | NP_001093979.1 | 1 - 2 | 141 | 138 | |
| GenBank mRNA | BC107950.1 | - | 1 - 2 | 62 | 138 | |
| DY570284.1 | - | 1 - 2 | 141 | 138 | ||
| FQ213949.1 | - | 1 - 2 | 89 | 138 | ||
| FQ214043.1 | - | 1 - 2 | 94 | 138 | ||
| FQ227742.1 | - | 1 - 2 | 98 | 138 | ||
Set Membership: |
|