Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01416971_g1
          See other DMAP1 GE Assays ›
          Gene Symbol:
          Dmap1
          Gene Name:
          DNA methyltransferase 1-associated protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 136533791 - 136541795 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01416971_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dmap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001015006.1 NP_001015006.1 9 - 10 1164 88
          GenBank mRNA BC092651.1 - 9 - 10 1164 7319534 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dmap1

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          DNA repair
          chromatin remodeling
          methylation
          positive regulation of transcription factor import into nucleus
          histone H4 acetylation
          histone H2A acetylation
          response to ethanol
          negative regulation of transcription, DNA-templated
          RNA polymerase II repressing transcription factor binding
          molecular_function
          chromatin binding
          transcription corepressor activity
          methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P550981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260457 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71682 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P550981 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P550981 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P550981 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P550981 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45511 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10879017 - -
          Affymetrix Rat Gene 1.0 ST Array 10879021 - -
          Affymetrix Rat Gene 1.0 ST Array 10938805 - -
          Affymetrix Rat Gene 1.0 ST Array 10879019 - -
          Affymetrix Rat Gene 1.0 ST Array 10879018 - -
          Affymetrix Rat Gene 1.0 ST Array 10879020 - -
          Affymetrix Rat Gene 1.0 ST Array 10879024 - -
          Affymetrix Rat Gene 1.0 ST Array 10879023 - -
          Affymetrix Rat Gene 1.0 ST Array 10879022 - -
          Affymetrix Rat Gene 1.1 ST Array 10938804 - -
          Affymetrix Rat Gene 1.1 ST Array 10879016 - -
          Rat Genome 230 2.0 Array 1389764_at - -
          Rat Genome 230 2.0 Array 1389764_PM_at - -
          Affymetrix HT Rat Focus Array 1389764_PM_at - -
          Affymetrix HT Rat Focus Array 1389764_at - -
          Affymetrix Rat Clariom S Assay PSR0500025244.rn.1 TC0500003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500025246.rn.1 TC0500003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500025243.rn.1 TC0500003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500025242.rn.1 TC0500003442.rn.1 RTA TC
          Rat Expression Set 230 1389764_at - -
          Rat Expression Set 230 1389764_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10879016 - -
          Affymetrix Rat Gene 1.0 ST Array 10938804 - -
          Rat Genome U34 Set rc_AA924980_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21448420 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline