Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01417086_m1
          See other KDM4A GE Assays ›
          Gene Symbol:
          Kdm4a
          Gene Name:
          lysine (K)-specific demethylase 4A
          Gene Aliases:
          Jmjd2a
          Chromosome Location:
          Chr.5: 136967650 - 137014375 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01417086_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kdm4a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107966.1 NP_001101436.1 17 - 18 2643 67
          XM_006238668.1 XP_006238730.1 18 - 19 2851 67
          XM_006238670.2 XP_006238732.1 16 - 17 2538 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003451.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of gene expression
          cardiac muscle hypertrophy in response to stress
          histone demethylation
          response to nutrient levels
          positive regulation of neuron differentiation
          negative regulation of transcription, DNA-templated
          negative regulation of astrocyte differentiation
          negative regulation of cell death
          histone H3-K36 demethylation
          double-stranded DNA binding
          zinc ion binding
          ubiquitin protein ligase binding
          histone demethylase activity
          methylated histone binding
          histone demethylase activity (H3-K36 specific)

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P199368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766305 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176863 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179792 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129295 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23195 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P199368 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P199368 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P199368 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P199368 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95712 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94383 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10879126 - -
          Affymetrix Rat Gene 1.0 ST Array 10879130 - -
          Affymetrix Rat Gene 1.0 ST Array 10879117 - -
          Affymetrix Rat Gene 1.0 ST Array 10879113 - -
          Affymetrix Rat Gene 1.0 ST Array 10879120 - -
          Affymetrix Rat Gene 1.0 ST Array 10879127 - -
          Affymetrix Rat Gene 1.0 ST Array 10879119 - -
          Affymetrix Rat Gene 1.0 ST Array 10879131 - -
          Affymetrix Rat Gene 1.0 ST Array 10879124 - -
          Affymetrix Rat Gene 1.0 ST Array 10879128 - -
          Affymetrix Rat Gene 1.0 ST Array 10879112 - -
          Affymetrix Rat Gene 1.0 ST Array 10879125 - -
          Affymetrix Rat Gene 1.0 ST Array 10879116 - -
          Affymetrix Rat Gene 1.0 ST Array 10879118 - -
          Affymetrix Rat Gene 1.0 ST Array 10879115 - -
          Affymetrix Rat Gene 1.0 ST Array 10879111 - -
          Affymetrix Rat Gene 1.0 ST Array 10879123 - -
          Affymetrix Rat Gene 1.0 ST Array 10879122 - -
          Affymetrix Rat Gene 1.0 ST Array 10879129 - -
          Affymetrix Rat Gene 1.0 ST Array 10879121 - -
          Affymetrix Rat Gene 1.1 ST Array 10879110 - -
          Affymetrix Rat Clariom S Assay PSR0500025431.rn.1 TC0500003451.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10879110 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20704958 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355854 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline