Search
Search
| Gene Symbol: Klhl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047093.1 | NP_001040558.1 | 5 - 6 | 1278 | 69 | |
| XM_006250959.2 | XP_006251021.1 | 5 - 6 | 934 | 69 | ||
| XM_008770157.1 | XP_008768379.1 | 5 - 6 | 756 | 69 | ||
| GenBank mRNA | AY641809.1 | - | 5 - 6 | 1278 | 69 | |
Set Membership: |
|