Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01418582_m1
          See other CCDC33 GE Assays ›
          Gene Symbol:
          Ccdc33
          Gene Name:
          coiled-coil domain containing 33
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 62811171 - 62893087 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01418582_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc33
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014091.1 NP_001014113.1 15 - 16 2050 97
          GenBank mRNA BC083609.1 - 15 - 16 2050 7133493 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003683.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc33

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1030081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14509 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66725 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172994 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86295 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57581 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177634 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70479 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131018 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23927 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1030081 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1030081 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1030081 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P356758 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1030081 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37717 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18548 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71620 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74760 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98267 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10917865 - -
          Affymetrix Rat Gene 1.0 ST Array 10917845 - -
          Affymetrix Rat Gene 1.0 ST Array 10917860 - -
          Affymetrix Rat Gene 1.0 ST Array 10917848 - -
          Affymetrix Rat Gene 1.0 ST Array 10917849 - -
          Affymetrix Rat Gene 1.0 ST Array 10917843 - -
          Affymetrix Rat Gene 1.0 ST Array 10917863 - -
          Affymetrix Rat Gene 1.0 ST Array 10917847 - -
          Affymetrix Rat Gene 1.0 ST Array 10917859 - -
          Affymetrix Rat Gene 1.0 ST Array 10917858 - -
          Affymetrix Rat Gene 1.0 ST Array 10917842 - -
          Affymetrix Rat Gene 1.0 ST Array 10917861 - -
          Affymetrix Rat Gene 1.0 ST Array 10917856 - -
          Affymetrix Rat Gene 1.0 ST Array 10917850 - -
          Affymetrix Rat Gene 1.0 ST Array 10917862 - -
          Affymetrix Rat Gene 1.0 ST Array 10917852 - -
          Affymetrix Rat Gene 1.0 ST Array 10917854 - -
          Affymetrix Rat Gene 1.0 ST Array 10917844 - -
          Affymetrix Rat Gene 1.0 ST Array 10917853 - -
          Affymetrix Rat Gene 1.0 ST Array 10917864 - -
          Affymetrix Rat Gene 1.0 ST Array 10917851 - -
          Affymetrix Rat Gene 1.0 ST Array 10917857 - -
          Affymetrix Rat Gene 1.0 ST Array 10917866 - -
          Affymetrix Rat Gene 1.1 ST Array 10917841 - -
          Affymetrix Rat Clariom S Assay PSR0800030395.rn.1 TC0800003683.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800030397.rn.1 TC0800003683.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800030398.rn.1 TC0800003683.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800030399.rn.1 TC0800003683.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800030394.rn.1 TC0800003683.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800030393.rn.1 TC0800003683.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800030400.rn.1 TC0800003683.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800030396.rn.1 TC0800003683.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10917841 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22242994 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362928 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline