Search
Search
| Gene Symbol: Bcl9l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106817.1 | NP_001100287.1 | 5 - 6 | 1326 | 58 | |
| XM_006242923.2 | XP_006242985.1 | 5 - 6 | 2888 | 58 | ||
| XM_006242924.2 | XP_006242986.1 | 4 - 5 | 2831 | 58 | ||
| XM_006242925.2 | XP_006242987.1 | 4 - 5 | 2786 | 58 | ||
| XM_006242928.2 | XP_006242990.1 | 2 - 3 | 511 | 58 | ||
Set Membership: |
|