Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01419341_m1
          See other RS1 GE Assays ›
          Gene Symbol:
          Rs1
          Gene Name:
          retinoschisin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 35749957 - 35777243 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01419341_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rs1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001104643.1 NP_001098113.1 5 - 6 519 128
          GenBank mRNA EU074986.1 - 5 - 6 519 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001674.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          retina layer formation
          adaptation of rhodopsin mediated signaling
          phosphatidylserine binding
          molecular_function
          phosphatidylinositol-4,5-bisphosphate binding
          phosphatidylinositol-3,4,5-trisphosphate binding
          phosphatidylethanolamine binding
          phosphatidylinositol-5-phosphate binding
          phosphatidylcholine binding
          phosphatidylinositol-3-phosphate binding
          phosphatidylinositol-3,4-bisphosphate binding
          phosphatidylinositol-4-phosphate binding
          phosphatidylinositol-3,5-bisphosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P234373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76400 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4026 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132747 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126693 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1732 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P234373 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P234373 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P234373 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4232 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10937863 - -
          Affymetrix Rat Gene 1.0 ST Array 10937865 - -
          Affymetrix Rat Gene 1.0 ST Array 10937866 - -
          Affymetrix Rat Gene 1.0 ST Array 10937864 - -
          Affymetrix Rat Gene 1.1 ST Array 10937862 - -
          Affymetrix Rat Clariom S Assay PSR0X00010845.rn.1 TC0X00001674.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10937862 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21403370 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline