Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01419801_m1
          See other DYDC2 GE Assays ›
          Gene Symbol:
          Dydc2
          Gene Name:
          DPY30 domain containing 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 18635234 - 18649074 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01419801_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dydc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134797.1 NP_001128269.1 5 - 6 542 95
          XM_006252813.2 XP_006252875.1 3 - 4 523 95
          XM_006252814.2 XP_006252876.1 4 - 5 434 95
          XM_006252816.2 XP_006252878.1 3 - 4 363 95
          XM_008771149.1 XP_008769371.1 3 - 4 386 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001323.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P040700 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P022950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149342 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50300 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040700 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022950 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040700 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022950 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P040700 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P022950 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10790649 - -
          Affymetrix Rat Gene 1.0 ST Array 10790648 - -
          Affymetrix Rat Gene 1.0 ST Array 10790650 - -
          Affymetrix Rat Gene 1.1 ST Array 10790647 - -
          Affymetrix Rat Clariom S Assay PSR1600009914.rn.1 TC1600001323.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009912.rn.1 TC1600001323.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009915.rn.1 TC1600001323.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009919.rn.1 TC1600001323.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009923.rn.1 TC1600001323.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009913.rn.1 TC1600001323.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10790647 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21274385 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline