Search
Search
| Gene Symbol: Akap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238184.2 | XP_006238246.1 | 5 | 107 | ||
| XM_006238185.2 | XP_006238247.1 | 5 | 107 | |||
| XM_006238186.2 | XP_006238248.1 | 5 | 107 | |||
| XM_008763733.1 | XP_008761955.1 | 6 | 107 | |||
Set Membership: |
|