Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01424867_g1
          See other ATF7IP2 GE Assays ›
          Gene Symbol:
          Atf7ip2
          Gene Name:
          activating transcription factor 7 interacting protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 5476205 - 5519938 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01424867_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atf7ip2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017471.1 NP_001017471.1 7 - 8 800 115
          XM_006245728.2 XP_006245790.1 7 - 8 954 115
          XM_006245732.2 XP_006245794.1 7 - 8 920 115
          XM_006245735.1 XP_006245797.1 5 - 6 528 115
          XM_008767495.1 XP_008765717.1 7 - 8 955 115
          XM_008767496.1 XP_008765718.1 7 - 8 953 115
          GenBank mRNA BC083663.1 - 7 - 8 800 7135436 115

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atf7ip2

          Panther Classification:

          Molecular Function -

          transcription cofactor

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P116774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656404 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51121 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42629 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6139 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85207 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54173 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116774 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116774 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P116774 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P173610 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9075 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63892 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95136 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10740480 - -
          Affymetrix Rat Gene 1.0 ST Array 10740486 - -
          Affymetrix Rat Gene 1.0 ST Array 10740490 - -
          Affymetrix Rat Gene 1.0 ST Array 10740485 - -
          Affymetrix Rat Gene 1.0 ST Array 10740481 - -
          Affymetrix Rat Gene 1.0 ST Array 10740495 - -
          Affymetrix Rat Gene 1.0 ST Array 10740487 - -
          Affymetrix Rat Gene 1.0 ST Array 10740491 - -
          Affymetrix Rat Gene 1.0 ST Array 10740482 - -
          Affymetrix Rat Gene 1.0 ST Array 10740489 - -
          Affymetrix Rat Gene 1.0 ST Array 10740488 - -
          Affymetrix Rat Gene 1.0 ST Array 10740494 - -
          Affymetrix Rat Gene 1.1 ST Array 10740479 - -
          Affymetrix Rat Clariom S Assay PSR1000019946.rn.1 TC1000002217.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000019945.rn.1 TC1000002217.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000019947.rn.1 TC1000002217.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000019948.rn.1 TC1000002217.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000019955.rn.1 TC1000002217.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000019956.rn.1 TC1000002217.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10740479 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21783824 - -
          ABI Rat Genome Survey Microarray 22187687 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355887 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline