Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01426354_m1
          See other FANCD2 GE Assays ›
          Gene Symbol:
          Fancd2
          Gene Name:
          Fanconi anemia, complementation group D2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 145489869 - 145551479 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01426354_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fancd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001719.1 NP_001001719.1 41 - 42 4188 135
          GenBank mRNA AY621075.1 - 41 - 42 4188 135

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001577.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fancd2

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          cellular response to DNA damage stimulus
          synapsis
          gamete generation
          biological_process
          response to gamma radiation
          molecular_function
          DNA polymerase binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P189375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469303 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32543 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165071 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117369 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8817 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10730 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62784 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94676 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P189375 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P189375 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P189375 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P189375 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24977 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36643 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79172 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81422 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88679 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66963 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10857856 - -
          Affymetrix Rat Gene 1.0 ST Array 10857877 - -
          Affymetrix Rat Gene 1.0 ST Array 10857858 - -
          Affymetrix Rat Gene 1.0 ST Array 10857851 - -
          Affymetrix Rat Gene 1.0 ST Array 10857882 - -
          Affymetrix Rat Gene 1.0 ST Array 10857871 - -
          Affymetrix Rat Gene 1.0 ST Array 10857845 - -
          Affymetrix Rat Gene 1.0 ST Array 10857842 - -
          Affymetrix Rat Gene 1.0 ST Array 10857879 - -
          Affymetrix Rat Gene 1.0 ST Array 10857844 - -
          Affymetrix Rat Gene 1.0 ST Array 10857865 - -
          Affymetrix Rat Gene 1.0 ST Array 10857870 - -
          Affymetrix Rat Gene 1.0 ST Array 10857843 - -
          Affymetrix Rat Gene 1.0 ST Array 10857840 - -
          Affymetrix Rat Gene 1.0 ST Array 10857859 - -
          Affymetrix Rat Gene 1.0 ST Array 10857841 - -
          Affymetrix Rat Gene 1.0 ST Array 10857875 - -
          Affymetrix Rat Gene 1.0 ST Array 10857867 - -
          Affymetrix Rat Gene 1.0 ST Array 10857874 - -
          Affymetrix Rat Gene 1.0 ST Array 10857855 - -
          Affymetrix Rat Gene 1.0 ST Array 10857878 - -
          Affymetrix Rat Gene 1.0 ST Array 10857868 - -
          Affymetrix Rat Gene 1.0 ST Array 10857839 - -
          Affymetrix Rat Gene 1.0 ST Array 10857846 - -
          Affymetrix Rat Gene 1.0 ST Array 10857847 - -
          Affymetrix Rat Gene 1.0 ST Array 10857848 - -
          Affymetrix Rat Gene 1.0 ST Array 10857861 - -
          Affymetrix Rat Gene 1.0 ST Array 10857863 - -
          Affymetrix Rat Gene 1.0 ST Array 10857872 - -
          Affymetrix Rat Gene 1.0 ST Array 10857862 - -
          Affymetrix Rat Gene 1.0 ST Array 10857860 - -
          Affymetrix Rat Gene 1.0 ST Array 10857866 - -
          Affymetrix Rat Gene 1.0 ST Array 10857853 - -
          Affymetrix Rat Gene 1.0 ST Array 10857849 - -
          Affymetrix Rat Gene 1.0 ST Array 10857869 - -
          Affymetrix Rat Gene 1.0 ST Array 10857873 - -
          Affymetrix Rat Gene 1.0 ST Array 10857857 - -
          Affymetrix Rat Gene 1.0 ST Array 10857876 - -
          Affymetrix Rat Gene 1.0 ST Array 10857854 - -
          Affymetrix Rat Gene 1.0 ST Array 10857852 - -
          Affymetrix Rat Gene 1.0 ST Array 10857850 - -
          Affymetrix Rat Gene 1.0 ST Array 10857864 - -
          Affymetrix Rat Gene 1.0 ST Array 10857880 - -
          Affymetrix Rat Gene 1.1 ST Array 10857838 - -
          Rat Genome 230 2.0 Array 1378240_at - -
          Rat Genome 230 2.0 Array 1378240_PM_at - -
          Affymetrix HT Rat Focus Array 1378240_at - -
          Affymetrix HT Rat Focus Array 1378240_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400011233.rn.1 TC0400001577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011235.rn.1 TC0400001577.rn.1 RTA TC
          Rat Expression Set 230 1378240_PM_at - -
          Rat Expression Set 230 1378240_at - -
          Affymetrix Rat Gene 1.0 ST Array 10857838 - -
          Rat Genome U34 Set rc_AI031007_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376860 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0