Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01426628_g1
          See other ACTA1 GE Assays ›
          Gene Symbol:
          Acta1
          Gene Name:
          actin, alpha 1, skeletal muscle
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 56674072 - 56677084 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01426628_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acta1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019212.2 NP_062085.1 3 - 4 512 87
          GenBank mRNA BC061974.1 - 3 - 4 512 5623909 87
          FQ214583.1 - 3 - 4 521 87
          FQ214585.1 - 3 - 4 523 87
          FQ214598.1 - 3 - 4 521 87
          FQ214600.1 - 3 - 4 521 87
          FQ214610.1 - 1 - 2 66 87
          FQ214623.1 - 3 - 4 521 87
          FQ214640.1 - 3 - 4 521 87
          FQ214659.1 - 3 - 4 523 87
          FQ214676.1 - 3 - 4 521 87
          FQ214680.1 - 3 - 4 521 87
          FQ214697.1 - 3 - 4 521 87
          FQ214734.1 - 3 - 4 521 87
          FQ214743.1 - 3 - 4 521 87
          FQ214756.1 - 3 - 4 521 87
          FQ214761.1 - 3 - 4 520 87
          FQ214779.1 - 3 - 4 521 87
          FQ214785.1 - 3 - 4 519 87
          FQ214807.1 - 3 - 4 521 87
          FQ214814.1 - 3 - 4 521 87
          FQ214822.1 - 3 - 4 521 87
          FQ214831.1 - 3 - 4 521 87
          FQ214832.1 - 3 - 4 521 87
          FQ214839.1 - 3 - 4 524 87
          FQ214841.1 - 3 - 4 521 87
          FQ214857.1 - 3 - 4 525 87
          FQ214858.1 - 3 - 4 521 87
          FQ214859.1 - 3 - 4 521 87
          FQ214860.1 - 3 - 4 521 87
          FQ214862.1 - 3 - 4 521 87
          FQ214866.1 - 3 - 4 521 87
          FQ214883.1 - 3 - 4 520 87
          FQ214904.1 - 3 - 4 521 87
          FQ214905.1 - 3 - 4 520 87
          FQ214916.1 - 3 - 4 521 87
          FQ214936.1 - 3 - 4 521 87
          FQ214948.1 - 3 - 4 521 87
          FQ214949.1 - 3 - 4 521 87
          FQ214969.1 - 3 - 4 521 87
          FQ214975.1 - 3 - 4 523 87
          FQ214976.1 - 3 - 4 521 87
          FQ214978.1 - 3 - 4 521 87
          FQ214987.1 - 3 - 4 87
          FQ214990.1 - 3 - 4 521 87
          FQ214993.1 - 3 - 4 521 87
          FQ215012.1 - 3 - 4 521 87
          FQ215025.1 - 3 - 4 521 87
          FQ215038.1 - 3 - 4 521 87
          FQ215044.1 - 3 - 4 521 87
          FQ215058.1 - 3 - 4 521 87
          FQ215089.1 - 3 - 4 521 87
          FQ215112.1 - 3 - 4 521 87
          FQ215128.1 - 3 - 4 523 87
          FQ215149.1 - 3 - 4 521 87
          FQ215153.1 - 3 - 4 521 87
          FQ215158.1 - 3 - 4 519 87
          FQ215173.1 - 3 - 4 521 87
          FQ215212.1 - 3 - 4 521 87
          FQ215215.1 - 3 - 4 521 87
          FQ215225.1 - 3 - 4 521 87
          FQ215241.1 - 3 - 4 521 87
          FQ215249.1 - 3 - 4 522 87
          FQ215253.1 - 3 - 4 521 87
          FQ215256.1 - 3 - 4 521 87
          FQ215275.1 - 3 - 4 521 87
          FQ215280.1 - 3 - 4 521 87
          FQ215282.1 - 3 - 4 521 87
          FQ215298.1 - 3 - 4 523 87
          FQ215324.1 - 3 - 4 521 87
          FQ215333.1 - 3 - 4 521 87
          FQ215342.1 - 3 - 4 521 87
          FQ215346.1 - 3 - 4 521 87
          FQ215366.1 - 3 - 4 521 87
          FQ215380.1 - 3 - 4 521 87
          FQ215381.1 - 3 - 4 521 87
          FQ215393.1 - 3 - 4 521 87
          FQ215396.1 - 3 - 4 521 87
          FQ215398.1 - 3 - 4 521 87
          FQ215402.1 - 3 - 4 521 87
          FQ215403.1 - 3 - 4 521 87
          FQ215405.1 - 3 - 4 521 87
          FQ215414.1 - 3 - 4 523 87
          FQ215424.1 - 3 - 4 521 87
          FQ215431.1 - 3 - 4 521 87
          FQ215433.1 - 3 - 4 520 87
          FQ215434.1 - 3 - 4 523 87
          FQ215438.1 - 3 - 4 521 87
          FQ215446.1 - 3 - 4 521 87
          FQ215449.1 - 3 - 4 523 87
          FQ215454.1 - 3 - 4 521 87
          FQ215464.1 - 3 - 4 521 87
          FQ215465.1 - 3 - 4 521 87
          FQ215468.1 - 3 - 4 521 87
          FQ215470.1 - 3 - 4 521 87
          FQ215474.1 - 3 - 4 521 87
          FQ215483.1 - 3 - 4 521 87
          FQ215488.1 - 3 - 4 521 87
          FQ215518.1 - 3 - 4 530 87
          FQ215523.1 - 3 - 4 521 87
          FQ215537.1 - 3 - 4 523 87
          FQ215554.1 - 3 - 4 521 87
          FQ215567.1 - 3 - 4 87
          FQ215572.1 - 3 - 4 521 87
          FQ215593.1 - 3 - 4 521 87
          FQ215618.1 - 3 - 4 521 87
          FQ215628.1 - 3 - 4 521 87
          FQ215631.1 - 1 - 2 135 87
          FQ215655.1 - 3 - 4 519 87
          FQ215659.1 - 3 - 4 520 87
          FQ215673.1 - 3 - 4 521 87
          FQ215674.1 - 3 - 4 521 87
          FQ215690.1 - 3 - 4 521 87
          FQ215692.1 - 3 - 4 521 87
          FQ215785.1 - 3 - 4 521 87
          FQ215790.1 - 3 - 4 523 87
          FQ215799.1 - 3 - 4 521 87
          FQ215813.1 - 3 - 4 521 87
          FQ215835.1 - 3 - 4 521 87
          FQ215836.1 - 3 - 4 521 87
          FQ215839.1 - 3 - 4 521 87
          FQ215840.1 - 3 - 4 521 87
          FQ215841.1 - 3 - 4 521 87
          FQ215851.1 - 3 - 4 521 87
          FQ215852.1 - 3 - 4 519 87
          FQ215864.1 - 3 - 4 522 87
          FQ215866.1 - 3 - 4 523 87
          FQ215888.1 - 3 - 4 520 87
          FQ215893.1 - 3 - 4 522 87
          FQ215896.1 - 3 - 4 521 87
          FQ215907.1 - 3 - 4 523 87
          FQ215924.1 - 1 - 2 213 87
          FQ215950.1 - 3 - 4 521 87
          FQ215964.1 - 3 - 4 521 87
          FQ215972.1 - 3 - 4 521 87
          FQ216016.1 - 3 - 4 521 87
          FQ216035.1 - 3 - 4 521 87
          FQ216043.1 - 3 - 4 521 87
          FQ216050.1 - 3 - 4 521 87
          FQ216066.1 - 3 - 4 519 87
          FQ216138.1 - 3 - 4 521 87
          FQ216139.1 - 3 - 4 521 87
          FQ216168.1 - 3 - 4 519 87
          FQ216196.1 - 3 - 4 521 87
          FQ216224.1 - 3 - 4 521 87
          FQ216246.1 - 3 - 4 521 87
          FQ216276.1 - 3 - 4 521 87
          FQ216278.1 - 3 - 4 523 87
          FQ216287.1 - 3 - 4 519 87
          FQ216291.1 - 3 - 4 521 87
          FQ216293.1 - 3 - 4 521 87
          FQ216305.1 - 3 - 4 521 87
          FQ216329.1 - 3 - 4 521 87
          FQ216333.1 - 3 - 4 521 87
          FQ216350.1 - 3 - 4 521 87
          FQ216352.1 - 3 - 4 521 87
          FQ216360.1 - 3 - 4 521 87
          FQ216369.1 - 3 - 4 523 87
          FQ216403.1 - 3 - 4 521 87
          FQ216430.1 - 3 - 4 521 87
          FQ216509.1 - 3 - 4 519 87
          FQ216586.1 - 3 - 4 521 87
          FQ216603.1 - 3 - 4 521 87
          FQ216606.1 - 3 - 4 521 87
          FQ216607.1 - 3 - 4 517 87
          FQ216610.1 - 3 - 4 519 87
          FQ216620.1 - 3 - 4 521 87
          FQ216635.1 - 3 - 4 519 87
          FQ216639.1 - 3 - 4 521 87
          FQ216644.1 - 3 - 4 521 87
          FQ216664.1 - 3 - 4 519 87
          FQ216675.1 - 3 - 4 521 87
          FQ216676.1 - 3 - 4 521 87
          FQ216692.1 - 3 - 4 521 87
          FQ216703.1 - 3 - 4 521 87
          FQ216713.1 - 3 - 4 521 87
          FQ216721.1 - 3 - 4 521 87
          FQ216725.1 - 3 - 4 517 87
          FQ216739.1 - 3 - 4 521 87
          FQ216762.1 - 3 - 4 519 87
          FQ216767.1 - 3 - 4 521 87
          FQ216776.1 - 3 - 4 521 87
          FQ216777.1 - 3 - 4 521 87
          FQ216779.1 - 3 - 4 523 87
          FQ216793.1 - 3 - 4 521 87
          FQ216801.1 - 3 - 4 521 87
          FQ216814.1 - 3 - 4 515 87
          FQ216819.1 - 3 - 4 519 87
          FQ216916.1 - 3 - 4 521 87
          FQ216918.1 - 1 - 2 164 87
          FQ216961.1 - 3 - 4 521 87
          FQ216994.1 - 3 - 4 521 87
          FQ217061.1 - 3 - 4 523 87
          FQ217073.1 - 3 - 4 524 87
          FQ217091.1 - 3 - 4 520 87
          FQ217114.1 - 3 - 4 521 87
          FQ217122.1 - 3 - 4 521 87
          FQ217167.1 - 3 - 4 521 87
          FQ217173.1 - 3 - 4 521 87
          FQ217188.1 - 3 - 4 521 87
          FQ217194.1 - 3 - 4 521 87
          FQ217273.1 - 3 - 4 520 87
          FQ217280.1 - 3 - 4 641 87
          FQ217327.1 - 3 - 4 521 87
          FQ217329.1 - 3 - 4 521 87
          FQ217347.1 - 3 - 4 521 87
          FQ217353.1 - 3 - 4 521 87
          FQ217367.1 - 3 - 4 521 87
          FQ217397.1 - 3 - 4 521 87
          FQ217441.1 - 3 - 4 521 87
          FQ217443.1 - 3 - 4 523 87
          FQ217475.1 - 3 - 4 521 87
          FQ217535.1 - 3 - 4 521 87
          FQ217572.1 - 3 - 4 521 87
          FQ217593.1 - 3 - 4 521 87
          FQ217602.1 - 3 - 4 521 87
          FQ217615.1 - 3 - 4 521 87
          FQ217629.1 - 3 - 4 521 87
          FQ217641.1 - 3 - 4 518 87
          FQ217762.1 - 3 - 4 521 87
          FQ217826.1 - 3 - 4 521 87
          FQ217884.1 - 3 - 4 519 87
          FQ217893.1 - 3 - 4 521 87
          FQ217905.1 - 3 - 4 521 87
          FQ217908.1 - 3 - 4 521 87
          FQ217919.1 - 3 - 4 521 87
          FQ217937.1 - 3 - 4 521 87
          FQ217969.1 - 3 - 4 523 87
          FQ217990.1 - 3 - 4 521 87
          FQ218019.1 - 3 - 4 521 87
          FQ218034.1 - 3 - 4 521 87
          FQ223563.1 - 3 - 4 521 87
          FQ223681.1 - 3 - 4 523 87
          FQ223686.1 - 3 - 4 521 87
          FQ223702.1 - 3 - 4 521 87
          FQ223727.1 - 1 - 2 219 87
          FQ223898.1 - 3 - 4 521 87
          FQ223914.1 - 3 - 4 521 87
          FQ223926.1 - 3 - 4 521 87
          FQ223929.1 - 3 - 4 521 87
          FQ224009.1 - 3 - 4 521 87
          FQ224082.1 - 3 - 4 521 87
          FQ224141.1 - 3 - 4 522 87
          FQ224167.1 - 3 - 4 521 87
          FQ224359.1 - 3 - 4 521 87
          FQ224377.1 - 3 - 4 521 87
          FQ224419.1 - 3 - 4 521 87
          FQ224552.1 - 3 - 4 521 87
          FQ224691.1 - 3 - 4 519 87
          FQ224777.1 - 3 - 4 521 87
          FQ224840.1 - 3 - 4 519 87
          FQ224842.1 - 3 - 4 521 87
          FQ224862.1 - 3 - 4 521 87
          FQ224869.1 - 3 - 4 521 87
          FQ224886.1 - 3 - 4 521 87
          FQ224890.1 - 3 - 4 519 87
          FQ224902.1 - 3 - 4 521 87
          FQ224929.1 - 3 - 4 519 87
          FQ224936.1 - 3 - 4 521 87
          FQ224942.1 - 3 - 4 523 87
          FQ224948.1 - 3 - 4 523 87
          FQ224962.1 - 3 - 4 521 87
          FQ224971.1 - 3 - 4 521 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001667.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acta1

          Panther Classification:

          Molecular Function -

          actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to mechanical stimulus
          response to extracellular stimulus
          response to lithium ion
          cell growth
          skeletal muscle thin filament assembly
          skeletal muscle fiber adaptation
          response to steroid hormone
          skeletal muscle fiber development
          ATP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1900013471.rn.1 TC1900001667.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013470.rn.1 TC1900001667.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline