Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01427181_m1
          See other PPIP5K2 GE Assays ›
          Gene Symbol:
          Ppip5k2
          Gene Name:
          diphosphoinositol pentakisphosphate kinase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 111232967 - 111249613 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01427181_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppip5k2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025775.1 NP_001020946.1 4 - 5 673 133
          GenBank mRNA BC097365.1 - 4 - 5 673 7443073 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002710.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppip5k2

          Panther Classification:

          Molecular Function -

          nucleotide kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          dephosphorylation
          acid phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P801044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650894 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24065 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133905 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P801044 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P801044 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P801044 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P801044 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47551 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21290 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14629 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10925731 - -
          Affymetrix Rat Gene 1.0 ST Array 10925728 - -
          Affymetrix Rat Gene 1.0 ST Array 10925726 - -
          Affymetrix Rat Gene 1.0 ST Array 10925732 - -
          Affymetrix Rat Gene 1.0 ST Array 10925730 - -
          Affymetrix Rat Gene 1.0 ST Array 10925733 - -
          Affymetrix Rat Gene 1.0 ST Array 10925727 - -
          Affymetrix Rat Gene 1.0 ST Array 10925729 - -
          Affymetrix Rat Gene 1.1 ST Array 10925725 - -
          Affymetrix Rat Clariom S Assay PSR0900022278.rn.1 TC0900002710.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900022280.rn.1 TC0900002710.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900022279.rn.1 TC0900002710.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900022286.rn.1 TC0900002710.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900022277.rn.1 TC0900002710.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900022284.rn.1 TC0900002710.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900022288.rn.1 TC0900002710.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900022275.rn.1 TC0900002710.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900022282.rn.1 TC0900002710.rn.1 RTA TC
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372684 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline