Search
Search
| Gene Symbol: Taf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008350.1 | NP_001008351.1 | 1 - 2 | 248 | 70 | |
| XM_006256191.2 | XP_006256253.1 | 2 - 3 | 1078 | 70 | ||
| XM_006256192.2 | XP_006256254.1 | 2 - 3 | 879 | 70 | ||
| XM_008772812.1 | XP_008771034.1 | 2 - 3 | 862 | 70 | ||
| GenBank mRNA | BC086421.1 | - | 1 - 2 | 248 | 7300732 | 70 |
Set Membership: |
|