Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01428814_m1
          See other PRMT2 GE Assays ›
          Gene Symbol:
          Prmt2
          Gene Name:
          protein arginine methyltransferase 2
          Gene Aliases:
          Hrmt1l1
          Chromosome Location:
          Chr.20: 13154506 - 13212613 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01428814_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prmt2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025144.1 NP_001020315.1 9 - 10 1232 68
          GenBank mRNA BC079112.1 - 9 - 10 1232 7125748 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000440.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prmt2

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          immune response
          biological_process
          histone methylation
          peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
          negative regulation of NF-kappaB transcription factor activity
          histone arginine methylation
          peptidyl-arginine N-methylation
          positive regulation of apoptotic process
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          developmental cell growth
          regulation of androgen receptor signaling pathway
          negative regulation of G1/S transition of mitotic cell cycle
          positive regulation of extrinsic apoptotic signaling pathway
          molecular_function
          transcription coactivator activity
          cytokine activity
          tumor necrosis factor receptor binding
          beta-catenin binding
          histone-arginine N-methyltransferase activity
          protein-arginine N-methyltransferase activity
          estrogen receptor binding
          progesterone receptor binding
          protein-arginine omega-N asymmetric methyltransferase activity
          histone methyltransferase activity
          protein homodimerization activity
          retinoic acid receptor binding
          peroxisome proliferator activated receptor binding
          thyroid hormone receptor binding
          androgen receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P831282 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P831282 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P831282 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P831282 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P831282 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P234970 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P258497 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10829602 - -
          Affymetrix Rat Gene 1.0 ST Array 10829598 - -
          Affymetrix Rat Gene 1.0 ST Array 10829594 - -
          Affymetrix Rat Gene 1.0 ST Array 10829593 - -
          Affymetrix Rat Gene 1.0 ST Array 10829599 - -
          Affymetrix Rat Gene 1.0 ST Array 10829596 - -
          Affymetrix Rat Gene 1.0 ST Array 10829601 - -
          Affymetrix Rat Gene 1.0 ST Array 10829597 - -
          Affymetrix Rat Gene 1.0 ST Array 10829592 - -
          Affymetrix Rat Gene 1.0 ST Array 10829600 - -
          Affymetrix Rat Gene 1.0 ST Array 10829595 - -
          Affymetrix Rat Gene 1.1 ST Array 10829591 - -
          Rat Genome 230 2.0 Array 1376025_at - -
          Rat Genome 230 2.0 Array 1376025_PM_at - -
          Affymetrix HT Rat Focus Array 1376025_PM_at - -
          Affymetrix HT Rat Focus Array 1376025_at - -
          Affymetrix Rat Clariom S Assay PSR2000004306.rn.1 TC2000000440.rn.1 RTA TC
          Rat Expression Set 230 1376025_PM_at - -
          Rat Expression Set 230 1376025_at - -
          Affymetrix Rat Gene 1.0 ST Array 10829591 - -
          Rat Genome U34 Set rc_AA925008_at - -
          Rat Genome U34 Set rc_AA945753_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20881376 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350340 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline