Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01431358_g1
          See other CFDP1 GE Assays ›
          Gene Symbol:
          Cfdp1
          Gene Name:
          craniofacial development protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 43989596 - 44078320 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01431358_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cfdp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199378.3 NP_955410.1 6 - 7 914 66
          GenBank mRNA AB125856.1 - 6 - 7 794 66
          BC067246.1 - 6 - 7 918 6889499 66
          FQ212332.1 - 948 66

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cfdp1

          Gene Ontology Categories:

          Function(s)

          cell adhesion
          multicellular organism development
          regulation of cell shape
          regulation of cell proliferation
          negative regulation of fibroblast apoptotic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P025178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737741 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174541 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109226 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58876 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129312 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139090 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90212 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70373 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129292 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46609 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180348 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169779 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142672 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47353 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73112 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99120 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025178 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025178 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P025178 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P499472 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8433 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93693 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82245 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 921 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27910 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10811202 - -
          Affymetrix Rat Gene 1.0 ST Array 10811198 - -
          Affymetrix Rat Gene 1.0 ST Array 10811197 - -
          Affymetrix Rat Gene 1.0 ST Array 10811199 - -
          Affymetrix Rat Gene 1.0 ST Array 10811200 - -
          Affymetrix Rat Gene 1.0 ST Array 10811201 - -
          Affymetrix Rat Gene 1.1 ST Array 10811196 - -
          Rat Genome 230 2.0 Array 1373824_at - -
          Rat Genome 230 2.0 Array 1395825_PM_at - -
          Rat Genome 230 2.0 Array 1395825_at - -
          Rat Genome 230 2.0 Array 1373824_PM_at - -
          Affymetrix HT Rat Focus Array 1373824_PM_at - -
          Affymetrix HT Rat Focus Array 1395825_at - -
          Affymetrix HT Rat Focus Array 1373824_at - -
          Affymetrix HT Rat Focus Array 1395825_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900012182.rn.1 TC1900001479.rn.1 RTA TC
          Rat Expression Set 230 1373824_at - -
          Rat Expression Set 230 1373824_PM_at - -
          Rat Expression Set 230 1395825_at - -
          Rat Expression Set 230 1395825_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10811196 - -
          Rat Genome U34 Set rc_AI228299_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20745049 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366130 - -
          RatRef-12 Expression BeadChip ILMN_54497 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline