Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01432162_m1
          See other CC2D1A GE Assays ›
          Gene Symbol:
          Cc2d1a
          Gene Name:
          coiled-coil and C2 domain containing 1A
          Gene Aliases:
          RGD1306108
          Chromosome Location:
          Chr.19: 25246912 - 25261966 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01432162_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cc2d1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013869.1 NP_001013891.1 10 - 11 1222 77
          XM_006255233.2 XP_006255295.1 11 - 12 1411 77
          XM_006255234.2 XP_006255296.1 11 - 12 1411 77
          XM_008772364.1 XP_008770586.1 10 - 11 1277 77
          GenBank mRNA BC081948.1 - 10 - 11 1222 7122187 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001186.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cc2d1a

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          signal transduction
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          signal transducer activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P018688 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P060832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548962 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44548 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38845 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060832 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018688 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018688 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060832 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P018688 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P060832 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P406195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28458 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98951 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806738 - -
          Affymetrix Rat Gene 1.0 ST Array 10806733 - -
          Affymetrix Rat Gene 1.0 ST Array 10806747 - -
          Affymetrix Rat Gene 1.0 ST Array 10806740 - -
          Affymetrix Rat Gene 1.0 ST Array 10806748 - -
          Affymetrix Rat Gene 1.0 ST Array 10806749 - -
          Affymetrix Rat Gene 1.0 ST Array 10806755 - -
          Affymetrix Rat Gene 1.0 ST Array 10806750 - -
          Affymetrix Rat Gene 1.0 ST Array 10806756 - -
          Affymetrix Rat Gene 1.0 ST Array 10806758 - -
          Affymetrix Rat Gene 1.0 ST Array 10806732 - -
          Affymetrix Rat Gene 1.0 ST Array 10806757 - -
          Affymetrix Rat Gene 1.0 ST Array 10806734 - -
          Affymetrix Rat Gene 1.0 ST Array 10806742 - -
          Affymetrix Rat Gene 1.0 ST Array 10806737 - -
          Affymetrix Rat Gene 1.0 ST Array 10806735 - -
          Affymetrix Rat Gene 1.0 ST Array 10806741 - -
          Affymetrix Rat Gene 1.0 ST Array 10806761 - -
          Affymetrix Rat Gene 1.0 ST Array 10806739 - -
          Affymetrix Rat Gene 1.0 ST Array 10806751 - -
          Affymetrix Rat Gene 1.0 ST Array 10806754 - -
          Affymetrix Rat Gene 1.0 ST Array 10806745 - -
          Affymetrix Rat Gene 1.0 ST Array 10806743 - -
          Affymetrix Rat Gene 1.0 ST Array 10806744 - -
          Affymetrix Rat Gene 1.0 ST Array 10806752 - -
          Affymetrix Rat Gene 1.0 ST Array 10806753 - -
          Affymetrix Rat Gene 1.0 ST Array 10806746 - -
          Affymetrix Rat Gene 1.0 ST Array 10806760 - -
          Affymetrix Rat Gene 1.1 ST Array 10806731 - -
          Rat Genome 230 2.0 Array 1389750_at - -
          Rat Genome 230 2.0 Array 1379399_PM_at - -
          Rat Genome 230 2.0 Array 1379399_at - -
          Rat Genome 230 2.0 Array 1389750_PM_at - -
          Affymetrix HT Rat Focus Array 1389750_PM_at - -
          Affymetrix HT Rat Focus Array 1389750_at - -
          Affymetrix HT Rat Focus Array 1379399_at - -
          Affymetrix HT Rat Focus Array 1379399_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900009595.rn.1 TC1900001186.rn.1 RTA TC
          Rat Expression Set 230 1389750_PM_at - -
          Rat Expression Set 230 1389750_at - -
          Rat Expression Set 230 1379399_PM_at - -
          Rat Expression Set 230 1379399_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806731 - -
          Rat Genome U34 Set rc_AI030999_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21315904 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650981 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline