Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01434103_m1
          See other PUS10 GE Assays ›
          Gene Symbol:
          Pus10
          Gene Name:
          pseudouridylate synthase 10
          Gene Aliases:
          RGD1306402
          Chromosome Location:
          Chr.14: 108412152 - 108479701 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01434103_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pus10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025278.1 NP_001020449.1 8 - 9 1131 65
          XM_008770428.1 XP_008768650.1 8 - 9 1075 65
          XM_008770429.1 XP_008768651.1 8 - 9 1075 65
          GenBank mRNA BC093611.1 - 8 - 9 1131 7307810 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001151.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pus10

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P060725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604312 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99920 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98073 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46554 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060725 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060725 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P060725 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70478 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34425 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73023 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10774584 - -
          Affymetrix Rat Gene 1.0 ST Array 10774578 - -
          Affymetrix Rat Gene 1.0 ST Array 10774580 - -
          Affymetrix Rat Gene 1.0 ST Array 10774583 - -
          Affymetrix Rat Gene 1.0 ST Array 10774586 - -
          Affymetrix Rat Gene 1.0 ST Array 10774581 - -
          Affymetrix Rat Gene 1.0 ST Array 10774585 - -
          Affymetrix Rat Gene 1.0 ST Array 10774582 - -
          Affymetrix Rat Gene 1.0 ST Array 10774579 - -
          Affymetrix Rat Gene 1.1 ST Array 10774577 - -
          Rat Genome 230 2.0 Array 1396692_at - -
          Rat Genome 230 2.0 Array 1390194_at - -
          Rat Genome 230 2.0 Array 1396692_PM_at - -
          Rat Genome 230 2.0 Array 1390194_PM_at - -
          Affymetrix HT Rat Focus Array 1396692_at - -
          Affymetrix HT Rat Focus Array 1396692_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400008357.rn.1 TC1400001151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400008356.rn.1 TC1400001151.rn.1 RTA TC
          Rat Expression Set 230 1396692_PM_at - -
          Rat Expression Set 230 1396692_at - -
          Affymetrix Rat Gene 1.0 ST Array 10774577 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374752 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline