Search
Search
| Gene Symbol: Grin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270602.1 | NP_001257531.1 | 9 - 10 | 1584 | 73 | |
| NM_001270603.1 | NP_001257532.1 | 9 - 10 | 1584 | 73 | ||
| NM_001270605.1 | NP_001257534.1 | 8 - 9 | 1521 | 73 | ||
| NM_001270606.1 | NP_001257535.1 | 9 - 10 | 1584 | 73 | ||
| NM_001270608.1 | NP_001257537.1 | 9 - 10 | 1584 | 73 | ||
| NM_001270610.1 | NP_001257539.1 | 8 - 9 | 1521 | 73 | ||
| NM_001287423.1 | NP_001274352.1 | 8 - 9 | 1521 | 73 | ||
| NM_017010.2 | NP_058706.1 | 8 - 9 | 1521 | 73 | ||
| XM_006233555.2 | XP_006233617.1 | 9 - 10 | 1590 | 73 | ||
| XM_006233556.2 | XP_006233618.1 | 8 - 9 | 1527 | 73 | ||
| GenBank mRNA | U08261.1 | - | 8 - 9 | 1432 | 73 | |
| U08262.1 | - | 8 - 9 | 1432 | 73 | ||
| U08263.1 | - | 9 - 10 | 1471 | 73 | ||
| U08264.1 | - | 9 - 10 | 1471 | 73 | ||
| U08265.1 | - | 8 - 9 | 1432 | 73 | ||
| U08266.1 | - | 9 - 10 | 1471 | 73 | ||
| U08267.1 | - | 8 - 9 | 1432 | 73 | ||
| U08268.1 | - | 9 - 10 | 1471 | 73 | ||
| U11418.1 | - | 8 - 9 | 1337 | 73 | ||
| X63255.1 | - | 8 - 9 | 1462 | 73 | ||
| X65227.1 | - | 9 - 10 | 1260 | 73 | ||
Set Membership: |
|