Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01439667_m1
          See other RGD1305807 GE Assays ›
          Gene Symbol:
          RGD1305807
          Gene Name:
          hypothetical LOC298077
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 61425821 - 61467653 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01439667_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1305807
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025276.1 NP_001020447.1 15 - 16 1434 93
          XM_006238038.2 XP_006238100.1 14 - 15 1547 93
          XM_006238039.1 XP_006238101.1 14 - 15 1351 93
          GenBank mRNA BC097368.1 - 15 - 16 1434 7443399 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000578.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RGD1305807

          Gene Ontology Categories:

          Function(s)

          retinol transport
          vitamin A import

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P340261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169669 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171093 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74154 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57481 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76175 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69881 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P340261 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P340261 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P340261 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P340261 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64574 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54186 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33161 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54950 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10868750 - -
          Affymetrix Rat Gene 1.0 ST Array 10868735 - -
          Affymetrix Rat Gene 1.0 ST Array 10868745 - -
          Affymetrix Rat Gene 1.0 ST Array 10868732 - -
          Affymetrix Rat Gene 1.0 ST Array 10868733 - -
          Affymetrix Rat Gene 1.0 ST Array 10868738 - -
          Affymetrix Rat Gene 1.0 ST Array 10868743 - -
          Affymetrix Rat Gene 1.0 ST Array 10868747 - -
          Affymetrix Rat Gene 1.0 ST Array 10868749 - -
          Affymetrix Rat Gene 1.0 ST Array 10868739 - -
          Affymetrix Rat Gene 1.0 ST Array 10868730 - -
          Affymetrix Rat Gene 1.0 ST Array 10868748 - -
          Affymetrix Rat Gene 1.0 ST Array 10868744 - -
          Affymetrix Rat Gene 1.0 ST Array 10868734 - -
          Affymetrix Rat Gene 1.0 ST Array 10868736 - -
          Affymetrix Rat Gene 1.0 ST Array 10868740 - -
          Affymetrix Rat Gene 1.0 ST Array 10868731 - -
          Affymetrix Rat Gene 1.0 ST Array 10868746 - -
          Affymetrix Rat Gene 1.0 ST Array 10868737 - -
          Affymetrix Rat Gene 1.1 ST Array 10868729 - -
          Rat Genome 230 2.0 Array 1382218_PM_at - -
          Rat Genome 230 2.0 Array 1382218_at - -
          Affymetrix HT Rat Focus Array 1382218_PM_at - -
          Affymetrix HT Rat Focus Array 1382218_at - -
          Affymetrix Rat Clariom S Assay PSR0500003773.rn.1 TC0500000578.rn.1 RTA TC
          Rat Expression Set 230 1382218_PM_at - -
          Rat Expression Set 230 1382218_at - -
          Affymetrix Rat Gene 1.0 ST Array 10868729 - -
          Rat Genome U34 Set rc_AI030706_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21010981 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351420 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline