Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01439719_m1
          See other DNAI1 GE Assays ›
          Gene Symbol:
          Dnai1
          Gene Name:
          dynein, axonemal, intermediate chain 1
          Gene Aliases:
          Dnaic1
          Chromosome Location:
          Chr.5: 57947796 - 58017985 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01439719_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnai1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024342.1 NP_001019513.1 19 - 20 2128 68
          GenBank mRNA BC083662.1 - 19 - 20 2128 7135392 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000495.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnai1

          Panther Classification:

          Molecular Function -

          microtubule or microtubule-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          epithelial cilium movement
          biological_process
          metabolic process
          motor activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P806532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144955 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9490 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106625 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42855 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54381 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157093 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73189 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98970 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P806532 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P806532 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P806532 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P806532 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56795 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12843 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72018 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75673 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10868364 - -
          Affymetrix Rat Gene 1.0 ST Array 10868373 - -
          Affymetrix Rat Gene 1.0 ST Array 10868379 - -
          Affymetrix Rat Gene 1.0 ST Array 10868368 - -
          Affymetrix Rat Gene 1.0 ST Array 10868375 - -
          Affymetrix Rat Gene 1.0 ST Array 10868376 - -
          Affymetrix Rat Gene 1.0 ST Array 10868369 - -
          Affymetrix Rat Gene 1.0 ST Array 10868380 - -
          Affymetrix Rat Gene 1.0 ST Array 10868381 - -
          Affymetrix Rat Gene 1.0 ST Array 10868383 - -
          Affymetrix Rat Gene 1.0 ST Array 10868367 - -
          Affymetrix Rat Gene 1.0 ST Array 10868372 - -
          Affymetrix Rat Gene 1.0 ST Array 10868374 - -
          Affymetrix Rat Gene 1.0 ST Array 10868371 - -
          Affymetrix Rat Gene 1.0 ST Array 10868378 - -
          Affymetrix Rat Gene 1.0 ST Array 10868365 - -
          Affymetrix Rat Gene 1.0 ST Array 10868366 - -
          Affymetrix Rat Gene 1.0 ST Array 10868377 - -
          Affymetrix Rat Gene 1.0 ST Array 10868382 - -
          Affymetrix Rat Gene 1.0 ST Array 10868363 - -
          Affymetrix Rat Gene 1.0 ST Array 10868370 - -
          Affymetrix Rat Gene 1.1 ST Array 10868362 - -
          Rat Genome 230 2.0 Array 1392279_PM_at - -
          Rat Genome 230 2.0 Array 1392279_at - -
          Affymetrix HT Rat Focus Array 1392279_PM_at - -
          Affymetrix HT Rat Focus Array 1392279_at - -
          Affymetrix Rat Clariom S Assay PSR0500002992.rn.1 TC0500000495.rn.1 RTA TC
          Rat Expression Set 230 1392279_at - -
          Rat Expression Set 230 1392279_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10868362 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20839583 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359284 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline