Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01441947_m1
          See other NSD1 GE Assays ›
          Gene Symbol:
          Nsd1
          Gene Name:
          nuclear receptor binding SET domain protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 9840859 - 9955394 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01441947_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nsd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107337.1 NP_001100807.1 8 - 9 4267 60
          XM_006253620.2 XP_006253682.1 8 - 9 4516 60
          XM_006253621.2 XP_006253683.1 8 - 9 4618 60
          XM_006253622.2 XP_006253684.1 8 - 9 4346 60
          XM_006253623.2 XP_006253685.1 8 - 9 4505 60
          XM_006253624.2 XP_006253686.1 8 - 9 4442 60
          XM_006253625.2 XP_006253687.1 8 - 9 4621 60
          XM_006253627.2 XP_006253689.1 8 - 9 4515 60
          XM_008771491.1 XP_008769713.1 8 - 9 4571 60
          XM_008771492.1 XP_008769714.1 7 - 8 3482 60
          XM_008771493.1 XP_008769715.1 7 - 8 3772 60
          XM_008771494.1 XP_008769716.1 8 - 9 4515 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001330.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          gastrulation with mouth forming second
          biological_process
          histone H3-K36 methylation
          histone methylation
          histone H4-K20 methylation
          histone lysine methylation
          positive regulation of transcription, DNA-templated
          molecular_function
          chromatin binding
          transcription cofactor activity
          transcription corepressor activity
          zinc ion binding
          histone-lysine N-methyltransferase activity
          estrogen receptor binding
          histone methyltransferase activity
          histone methyltransferase activity (H4-K20 specific)
          retinoic acid receptor binding
          retinoid X receptor binding
          thyroid hormone receptor binding
          histone methyltransferase activity (H3-K36 specific)
          androgen receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P037205 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P079796 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P21528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182791 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21855 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45165 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33983 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121104 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56909 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62324 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118695 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152816 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112943 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82089 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43946 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81937 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100011 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21528 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079796 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P037205 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P037205 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21528 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079796 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P21528 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P037205 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P079796 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P527432 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20192 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P22823 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54332 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96495 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70043 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60193 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797311 - -
          Affymetrix Rat Gene 1.0 ST Array 10797313 - -
          Affymetrix Rat Gene 1.0 ST Array 10797305 - -
          Affymetrix Rat Gene 1.0 ST Array 10797306 - -
          Affymetrix Rat Gene 1.0 ST Array 10797299 - -
          Affymetrix Rat Gene 1.0 ST Array 10797316 - -
          Affymetrix Rat Gene 1.0 ST Array 10797317 - -
          Affymetrix Rat Gene 1.0 ST Array 10797310 - -
          Affymetrix Rat Gene 1.0 ST Array 10797307 - -
          Affymetrix Rat Gene 1.0 ST Array 10797300 - -
          Affymetrix Rat Gene 1.0 ST Array 10797309 - -
          Affymetrix Rat Gene 1.0 ST Array 10797312 - -
          Affymetrix Rat Gene 1.0 ST Array 10797315 - -
          Affymetrix Rat Gene 1.0 ST Array 10797320 - -
          Affymetrix Rat Gene 1.0 ST Array 10797314 - -
          Affymetrix Rat Gene 1.0 ST Array 10797303 - -
          Affymetrix Rat Gene 1.0 ST Array 10797304 - -
          Affymetrix Rat Gene 1.0 ST Array 10797302 - -
          Affymetrix Rat Gene 1.0 ST Array 10797308 - -
          Affymetrix Rat Gene 1.0 ST Array 10797301 - -
          Affymetrix Rat Gene 1.0 ST Array 10797318 - -
          Affymetrix Rat Gene 1.1 ST Array 10797298 - -
          Rat Genome 230 2.0 Array 1380221_PM_at - -
          Rat Genome 230 2.0 Array 1398956_PM_at - -
          Rat Genome 230 2.0 Array 1380221_at - -
          Rat Genome 230 2.0 Array 1379560_at - -
          Rat Genome 230 2.0 Array 1379560_PM_at - -
          Rat Genome 230 2.0 Array 1398956_at - -
          Affymetrix HT Rat Focus Array 1379560_at - -
          Affymetrix HT Rat Focus Array 1379560_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700009215.rn.1 TC1700001330.rn.1 RTA TC
          Rat Expression Set 230 1398956_at - -
          Rat Expression Set 230 1398956_PM_at - -
          Rat Expression Set 230 1380221_at - -
          Rat Expression Set 230 1379560_PM_at - -
          Rat Expression Set 230 1380221_PM_at - -
          Rat Expression Set 230 1379560_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797298 - -
          Rat Genome U34 Set rc_AI058963_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21440898 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354380 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline